Skip to Content
Merck
All Photos(1)

Key Documents

SRP0313

Sigma-Aldrich

FBXL10 human

recombinant, expressed in baculovirus infected Sf9 cells, ≥80% (SDS-PAGE)

Synonym(s):

CXXC2, FBXL10, JHDM1B, KDM2B, lysine (K)-specific demethylase 2B

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
€713.00

€713.00


Usually ships in 2 weeks.


Select a Size

Change View
20 μG
€713.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

€713.00


Usually ships in 2 weeks.

biological source

human

recombinant

expressed in baculovirus infected Sf9 cells

Assay

≥80% (SDS-PAGE)

form

aqueous solution

mol wt

75 kDa

packaging

pkg of 20 μg

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... KDM2B(84678)

General description

Human F-Box Protein 10 (FBXL10, also known as KDM2B, JHDM1B, CXXC2, and Histone-H3 K36 demethylase 1B), (GenBank Accession No. NM_032590), amino acids 1-650 with C-terminal FLAG-tag, MW= 75 kDa, expressed in Sf9 cells via a Baculovirus expression system.

Application

Useful for the study of enzyme kinetics, screening inhibitors, and selectivity profiling.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Neil P Blackledge et al.
Molecular cell, 38(2), 179-190 (2010-04-27)
In higher eukaryotes, up to 70% of genes have high levels of nonmethylated cytosine/guanine base pairs (CpGs) surrounding promoters and gene regulatory units. These features, called CpG islands, were identified over 20 years ago, but there remains little mechanistic evidence
Yuji Tanaka et al.
The EMBO journal, 29(9), 1510-1522 (2010-04-10)
The rate-limiting step in ribosome biogenesis is the transcription of ribosomal RNA, which is controlled by environmental conditions. The JmjC enzyme KDM2A/JHDM1A/FbxL11 demethylates mono- and dimethylated Lys 36 of histone H3, but its function is unclear. Here, we show that
David Frescas et al.
Cell cycle (Georgetown, Tex.), 7(22), 3539-3547 (2008-11-13)
Heterochromatin plays an essential role in the preservation of epigenetic information, the transcriptional repression of repetitive DNA elements and inactive genes, and the proper segregation of chromosomes during mitosis. Here we identify KDM2A, a JmjC-domain containing histone demethylase, as a

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service