Skip to Content
Merck
All Photos(2)

Key Documents

SRP0228

Sigma-Aldrich

SMURF2 Active human

recombinant, expressed in baculovirus infected insect cells, ≥80% (SDS-PAGE)

Synonym(s):

E3 ubiquitin ligase SMURF2, MGC138150, SMAD specific E3 ubiquitin protein ligase 2

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
€557.00

€557.00


Usually ships in 2 weeks.

Request a Bulk Order

Select a Size

Change View
20 μG
€557.00

About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

€557.00


Usually ships in 2 weeks.

Request a Bulk Order

biological source

human

recombinant

expressed in baculovirus infected insect cells

Assay

≥80% (SDS-PAGE)

form

aqueous solution

specific activity

≥0.00049 pmol/min-μg

mol wt

72 kDa

packaging

pkg of 20 μg

storage condition

avoid repeated freeze/thaw cycles

concentration

>0.02 mg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... SMURF2(64750)

General description

Human SMURF 2 (GenBank Accession No. NM_022739), amino acids 130-end, with N-terminal DDDDK tag (FLAG), MW = 72 kDa, expressed in a Baculovirus infected Sf9 cell expression system.

Application

Useful in conjunction with E1 and E2 for the study of enzyme kinetics, screening inhibitors, and selectivity profiling.

Unit Definition

One unit is defined as the amount of enzyme required to transfer 1 pmol of ubiquitin to substrate/min at 37°C.

Physical form

Formulated in 25 mM Tris-HCl, pH 8.0, 100 mM NaCl, 0.05% Tween-20, 100 μg/ml FLAG peptide, 20% glycerol and 3 mM DTT.

Preparation Note

Thaw on ice. Upon first thaw, briefly spin tube containing enzyme to recover full content of the tube. Aliquot enzyme into single use aliquots. Store remaining undiluted enzyme in aliquots at -70°C. Note: Enzyme is very sensitive to freeze/thaw cycles.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Paramita Ray et al.
The Journal of biological chemistry, 295(36), 12661-12673 (2020-07-17)
The discovery of activating epidermal growth factor receptor (EGFR) mutations spurred the use of EGFR tyrosine kinase inhibitors (TKIs), such as erlotinib, as the first-line treatment of lung cancers. We previously reported that differential degradation of TKI-sensitive (e.g. L858R) and
LncRNA LITATS1 suppresses TGF-I?-induced EMT and cancer cell plasticity by potentiating TI?RI degradation.
Fan, et al.
The Embo Journal, 42, e112806-e112806 (2023)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service