Skip to Content
Merck
All Photos(1)

Key Documents

SRP0200

Sigma-Aldrich

Peroxiredoxin I (K197Q) human

recombinant, expressed in baculovirus infected insect cells, ≥80% (SDS-PAGE)

Synonym(s):

NKEF-A, Natural killer cell-enhancing factor A, Proliferation-associated gene A (PAGA), TDPX2, Thioredoxin peroxidase 2

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€690.00

€690.00


Usually ships in 2 weeks.

Request a Bulk Order

Select a Size

Change View
100 μG
€690.00

About This Item

MDL number:
UNSPSC Code:
12352200
NACRES:
NA.32

€690.00


Usually ships in 2 weeks.

Request a Bulk Order

biological source

human

recombinant

expressed in baculovirus infected insect cells

Assay

≥80% (SDS-PAGE)

form

aqueous solution

mol wt

23 kDa

packaging

pkg of 100 μg

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... PRDX1(5052)

General description

Human Peroxiredoxin I (GenBank Accession No. NM_005614), the full length protein containing a Lysine to Glutamine mutation at position 197 and N-terminal His6 tag, MW = 23 kDa, expressed in a Baculovirus infected Sf9 cell expression system.

Application

Useful for the study of biochemical function.

Physical form

Formulated in 25 mM Tris-HCl, pH 8.0, 100 mM NaCl, 0.05% Tween-20, 50% glycerol and 3 mM DTT.

Preparation Note

Thaw on ice. Upon first thaw, briefly spin tube containing enzyme to recover full content of the tube. Aliquot enzyme into single use aliquots. Store remaining undiluted enzyme in aliquots at -70°C. Note: Enzyme is very sensitive to freeze/thaw cycles.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service