Skip to Content
Merck
All Photos(1)

Key Documents

SRP0138

Sigma-Aldrich

NSD2 (782-end)/ReBPII Active human

recombinant, expressed in baculovirus infected insect cells, ≥70% (SDS-PAGE)

Synonym(s):

MMSET, Multiple myeloma SET domain-containing protein, Nuclear SET domain-containing protein 2, REIIBP, Wolf-Hirschhorn syndrome candidate 1

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
€673.00

€673.00


Usually ships in 2 weeks.


Select a Size

Change View
20 μG
€673.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

€673.00


Usually ships in 2 weeks.

biological source

human

recombinant

expressed in baculovirus infected insect cells

Assay

≥70% (SDS-PAGE)

form

aqueous solution

mol wt

66.5 kDa

packaging

pkg of 20 μg

concentration

>0.02 mg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... WHSC1(7468)

General description

Human NSD2 (GenBank Accession No. NM_133330), amino acids 782-end with N-terminal DDDDK (FLAG)-His tag, MW = 66.5 kDa, expressed in Baculovirus infected Sf9 cell expression system.

Application

Useful for the study of enzyme kinetics, screening inhibitors, and selectivity profiling.

Physical form

Formulated in 25 mM Tris-HCl, pH 8.0, 100 mM NaCl, 0.05% Tween-20, 20% glycerol, 100 μg/ml FLAG peptide and 3 mM DTT.

Preparation Note

Thaw on ice. Upon first thaw, briefly spin tube containing enzyme to recover full content of the tube. Aliquot enzyme into single use aliquots. Store remaining undiluted enzyme in aliquots at -70°C. Note: Enzyme is very sensitive to freeze/thaw cycles.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service