Skip to Content
Merck
All Photos(1)

Key Documents

SRE0021

Sigma-Aldrich

Granzyme B human

recombinant, expressed in insect cells, ≥90% (SDS-PAGE)

Synonym(s):

Granzyme B human, Cytotoxic T-lymphocyte proteinase 2, Granzyme-2, GranzymeB, Lymphocyte protease, SECT, T-cell serine protease 1-3E

Sign Into View Organizational & Contract Pricing

Select a Size

5 μG
€497.00

€497.00


Usually ships in 2 weeks.

Request a Bulk Order

Select a Size

Change View
5 μG
€497.00

About This Item

UNSPSC Code:
12352204
NACRES:
NA.32

€497.00


Usually ships in 2 weeks.

Request a Bulk Order

biological source

human

Quality Level

recombinant

expressed in insect cells

Assay

≥90% (SDS-PAGE)

form

lyophilized powder

potency

>0.004 units/mg

specific activity

≥40 units/μg

packaging

pkg of 5 μg

UniProt accession no.

shipped in

wet ice

storage temp.

−70°C

Gene Information

human ... GZMB(3002)

Looking for similar products? Visit Product Comparison Guide

General description

Granzyme B (GzmB) is a serine protease which is stored in granules of activated cytotoxic T cells and NK cells. GZMB codes for granzyme B.[1]

Biochem/physiol Actions

Granzyme B is considered as the marker of activated CTLs (cytotoxic T-cells), which mediates target-cell apoptosis.[1] It induces apoptosis in caspase-dependent and caspase-independent manner.[2] After contact with target cells, granzyme B is directionally exocytosed, and then enters the target cell with assistance from perforin. Granzyme B specifically cleaves after Asp residues, and particularly processes and activates various pro-caspases. This induces apoptosis in the target cell.

Unit Definition

One unit is defined as the amount of enzyme that hydrolyzes 1 pmol/min of IETD-AFC per min at 37°C in 50 mM HEPES, pH 7.5 containing 10 mM CaCl2.

Physical form

Lyophilized powder, containing HEPES buffer (original solution at pH 7.5)

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Granzyme B enters the mitochondria in a Sam50-, Tim22-and mtHsp70-dependent manner to induce apoptosis
Chiusolo V, et al.
Cell Death and Differentiation, 24(4), 747-747 (2017)
Risk of generalized vitiligo is associated with the common 55R-94A-247H variant haplotype of GZMB (encoding granzyme B).
Ferrara TM, et al.
The Journal of Investigative Dermatology, 133(6), 1677-1677 (2013)

Questions

1–2 of 2 Questions  
  1. What is the molecular weight? 

    1 answer
    1. Granzyme B human recombinant, expressed in insect cells, Molecular weight is 32kDa as per Uniprot P10144. Please see the link below for more information:
      https://www.uniprot.org/uniprot/P10144

      Helpful?

  2. To whom it may concern, we have recently purchased the granzyme B (SRE0021) and its substrate (SCP0158). Is the enzyme in its active form? What are the best condition for resuspension? Thank you.

    1 answer
    1. The product is provided in active form. It does not require activation. Use 50 µl sterile deionized water to dissolve 5 µg protein to a final concentration of 0.1 µg protein/µl. This solution is stable for 1 year at –70 °C.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service