Skip to Content
Merck
All Photos(1)

Key Documents

SRE0008

Sigma-Aldrich

Galactose-1-phosphate Uridyltransferase from galactose-adapted yeast

Suitable for manufacturing of diagnostic kits and reagents, lyophilized powder, 20-60 units/mg protein (modified Warburg-Christian)

Synonym(s):

GALT, UDP glucose:α-D-galactose-1-phosphate uridyltransferase

Sign Into View Organizational & Contract Pricing

Select a Size

40 UNITS
€1,610.00
50 UNITS
€2,180.00

€1,610.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
40 UNITS
€1,610.00
50 UNITS
€2,180.00

About This Item

CAS Number:
Enzyme Commission number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.32

€1,610.00


Check Cart for Availability

Request a Bulk Order

Quality Level

form

lyophilized powder

specific activity

20-60 units/mg protein (modified Warburg-Christian)

composition

Protein, 15-35%

application(s)

diagnostic assay manufacturing

foreign activity

6-phosphogluconate dehydrogenase ≤0.5%
UDP glucose pyrophosphorylase and galactokinase ≤0.2%

shipped in

wet ice

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Related Categories

Biochem/physiol Actions

Galactose-1-phosphate uridyltransferase catalyzes the conversion of UDP-glucose to galactose-1-phosphate and galactose-1-phosphate to UDP-glucose.

Unit Definition

One unit will form 1.0 μmole of glucose 1-phosphate from UDP-glucose, galactose 1-phosphate and NADP+ per min at pH 8.7 at 25 °C as detected by a coupled system using phosphoglucomutase.

Physical form

Contains buffer salts as citrate and reduced glutathione

Analysis Note

Protein determined by modified Warburg-Christian

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service