Skip to Content
Merck
All Photos(1)

Key Documents

SML3994

Sigma-Aldrich

BODIPY TR-X NHS Ester

new

≥95% (HPLC)

Synonym(s):

(T-4)-[N-[6-[(2,5-Dioxo-1-pyrrolidinyl)oxy]-6-oxohexyl]-2-[4-[5-[[5-(2-thienyl)-2H-pyrrol-2-ylidene-κN]methyl]-1H-pyrrol-2-yl-κN]phenoxy]acetamidato]difluoroboron, 6-(((4-(4,4-Difluoro-5-(2-thienyl)-4-bora-3a,4a-diaza-s-indacene-3-yl)phenoxy)acetyl)amino)hexanoic acid succinimidyl ester, BODIPY TR-X, BODIPY TR-X NHS, BODIPY TR-X, SE

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C31H29BF2N4O6S
CAS Number:
Molecular Weight:
634.46
UNSPSC Code:
12352200

Quality Level

Assay

≥95% (HPLC)

form

powder

color

, Red To very dark purple

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

SMILES string

O=C(CCCCCNC(COC1=CC=C(C2=CC=C3C=C4C=CC(C5=CC=CS5)=[N]4[B+3]([F-])([N-]32)[F-])C=C1)=O)ON6C(CCC6=O)=O

Biochem/physiol Actions

Amine reactive, red luminous fluorescent labeling reagent that can be used to tag antibodies, proteins, peptides, and nucleic acids.

BODIPY TR-X NHS Ester is a versatile, amine-sensitive red fluorescent labeling reagent useful to tag antibodies, proteins, peptides, and nucleic acids. BODIPY TR dyes, also called boron-dipyrromethene tetramethylrhodamine, are known for their exceptional photophysical properties, including high fluorescence quality and photostability, with wide spectral tuning. BODIPY TR has a narrow absorption range (~ 500-550 nm) and a broader emission range (~ 580-640 nm) for multi-color cell labeling. BODIPY TR-X offers an extended excited-state lifetime for fluorescence polarization assays and significant two-photon excitation capability. BODIPY TR-X NHS Ester is a valuable tool for fluorescence microscopy, flow cytometry, and IHC applications.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Łukasz Bujak et al.
Nanoscale, 9(4), 1511-1519 (2017-01-10)
Förster resonant energy transfer (FRET) is a nonradiative process by which the energy of light absorbed by a donor molecule is transferred to an acceptor molecule over a distance of several nanometers. FRET plays a crucial role in photosynthesis and
Karen Martin et al.
Proteomics, 2(5), 499-512 (2002-05-03)
A two-color fluorescence detection method is described based upon covalently coupling the succinimidyl ester of BODIPY TR-X dye to proteins immobilized on polyvinylidene difluoride membranes, followed by detection of target proteins using the fluorogenic, precipitating substrate ELF 39-phosphate in combination
Highly potent and selective fluorescent antagonists of the human adenosine A? receptor based on the 1,2,4-triazolo[4,3-a]quinoxalin-1-one scaffold
Journal of Medicinal Chemistry, 55(4), 1771-1782 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service