Skip to Content
Merck
All Photos(1)

Key Documents

SML3989

Sigma-Aldrich

DRP1 Inhibitor, MIDI

new

≥98% (HPLC)

Synonym(s):

1,1′-(1,4-Phenylene)bis[3-(3-pyridinyl)-2-propen-1-one], MIDI

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H16N2O2
CAS Number:
Molecular Weight:
340.37
UNSPSC Code:
12352200

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 0.5 mg/mL, clear (Warmed)

storage temp.

room temp

SMILES string

O=C(/C=C/C1=CC=CN=C1)C2=CC=C(C(/C=C/C3=CC=CN=C3)=O)C=C2

Biochem/physiol Actions

Mitochondrial fission inhibitor that blocks DRP1 recruitment to mitochondria.



DRP1 Inhibitor MIDI is a cell-permeable, symmetrical pyridinyl-propenone compound that acts as a potent, time-dependent mitochondrial fission inhibitor (0.5 - 2 µM) and functionally mimics DRP1 knockout. MIDI is shown to covalently interact with DRP1-C367 and disrupt DRP1 interaction with its mitochondrial receptors MFF and MiD49/51. Blocks stress-induced mitochondrial fragmentation and promotes mitochondrial hyperfusion. MIDI does neither obstruct DRP1 tetramerization nor its GTPase activity. Poorly affects mitochondrial oxygen consumption rate and glycolysis activity.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chemical inhibition of mitochondrial fission via targeting the DRP1-receptor interaction
Cell Chemical Biology, 30(3) (2023)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service