Skip to Content
Merck
All Photos(1)

Key Documents

SML3897

Sigma-Aldrich

Tyramide alkyne

≥95% (HPLC)

Synonym(s):

AP1, Alkyne tyramide, Alkyne-phenol 1, N-[2-(4-Hydroxyphenyl)ethyl]-5-hexynamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H17NO2
CAS Number:
Molecular Weight:
231.29
UNSPSC Code:
12352200
NACRES:
NA.21
Pricing and availability is not currently available.

Quality Level

Assay

≥95% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

Biochem/physiol Actions

Cell penetrant and highly efficient clickable ascorbate peroxidase 2 (APEX2) substrate that significantly improves APEX-labeling efficiency in intact yeast cells
Tyramide alkyne (Alkyne-phenol 1) is a cell penetrant and highly efficient clickable ascorbate peroxidase 2 (APEX2) substrate that significantly improves APEX-labeling efficiency in intact yeast cells. Tyramide alkyne enable biotin-independent detection of labeled proteins.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dynamic mapping of proteome trafficking within and between living cells by TransitID
Cell, 186(15), 3307-3324 (2023)
Compartment-Specific Labeling of Bacterial Periplasmic Proteins by Peroxidase-Mediated Biotinylation
ACS infectious diseases, 4(6), 918-925 (2018)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service