Skip to Content
Merck
All Photos(1)

Documents

SML3474

Sigma-Aldrich

PF-06928215, cis-racemate

≥98% (HPLC)

Synonym(s):

Cis-2-(7-Oxo-5-phenyl-4,7-dihydropyrazolo[1,5-a]pyrimidine-3 carboxamido) cyclohexane-1-carboxylic acid, Cis-2-(7-Oxo-5-phenyl-4,7-dihydropyrazolo[1,5-a]pyrimidine-3-carboxamido)cyclohexanecarboxylic acid, PF-06928215 cis-enantiomers (1R,2S) and (1S,2R)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H20N4O4
CAS Number:
Molecular Weight:
380.40
MDL number:
UNSPSC Code:
51111800
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear (Warmed)

storage temp.

2-8°C

SMILES string

OC([C@H](CCCC1)[C@H]1NC(C2=C3N(N=C2)C(C=C(C4=CC=CC=C4)N3)=O)=O)=O

Biochem/physiol Actions

PF-06928215 is a high affinity (KD of the (1R,2S) enantiomer = 200 nM) active site-targeting, substrate-competitive cyclic GMP-AMP synthase (cGAS) inhibitor (IC50 = 4.9 μM using the (1R,2S) enantiomer; in the presence of 1 mM ATP, 0.3 mM GTP, 100 nM 45-bp interferon-stimulatory dsDNA (ISD)). PF-06928215 displays no inhibitory potency against dsDNA-induced IFN-β expression in cellular cGAS assays, most likely due to limited cell-permeability and/or high levels of cellular ATP and GTP in the reporter cells employed. Note: this product contains the two cis-enantiomers (1R,2S) and (1S,2R).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Discovery of PF-06928215 as a high affinity inhibitor of cGAS enabled by a novel fluorescence polarization assay
PLoS ONE, 12 (2017)
Wenfeng Zhao et al.
Journal of chemical information and modeling, 60(6), 3265-3276 (2020-05-28)
Cyclic GMP-AMP synthase (cGAS) has been recently uncovered to be a promising therapeutic target for immune-associated diseases. Until now, only a few inhibitors have been identified through high-throughput screening campaigns. Here, we reported the discovery of novel inhibitors for the
Double knockout of Akt2 and AMPK accentuates high fat diet-induced cardiac anomalies through a cGAS-STING-mediated mechanism
Biochimica et Biophysica Acta (BBA)-Molecular Basis of Disease, 1866(10), 165855-165855 (2020)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service