Skip to Content
Merck

Skip To

SML3402

HSip-1 trifluoroacetate hydrate

≥98% (HPLC)

Synonym(s):

Cyclen-AF-Cu trifluoroacetate hydrate, HSip 1, HSip1, Hydrogen Sulfide imaging probe-1, [N-(3′,6′-Dihydroxy-3-oxospiro[isobenzofuran-1(3H),9′-[9H]xanthen]-5-yl)-1,4,7,10-tetraazacyclododecane-1-acetamide-κN1,κN4,κN7,κN10]-copper(2+) trifluoroacetate hydrate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C30H33CuN5O6 · xC2HF3O2 · yH2O
Molecular Weight:
623.16 (anhydrous free base basis)
UNSPSC Code:
41121706
NACRES:
NA.21
Assay:
≥98% (HPLC)
Form:
powder
Storage condition:
protect from light
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

storage condition

protect from light

color

green to black

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C1OC2(C3=CC=C(NC(C[N]45CC[N@@]6([H])CC[N]7([H])CC[N@]([Cu@+2]765)([H])CC4)=O)C=C13)C8=CC=C(C=C8OC9=CC(O)=CC=C92)O.[y H2O]

Biochem/physiol Actions

HSip-1 (Cyclen-AF-Cu; Hydrogen Sulfide imaging probe-1) is a sensitive hydrogen sulfide (H2S) fluorescent probe that displays a selective turn-on fluorescence response to sulfide anion (Ex/Em = 491/516 nm), but not GSH, L-Cys, DL-Hcy, 2-ME, DTT, SCN-, Na2SO3, Na2S2O3, or sodium ascorbate. HSip-1 is not membrane-permeable and is commonly employed for detecting H2S in media/buffers, cell lysates, blood, physiological fluids and tissue homogenates. HSip-1 is commonly used in the range from 1-100 μM depending on the H2S concentrations in samples, optimal probe concentration needs to be titred.
Sensitive and selective hydrogen sulfide (H2S) fluorescent probe.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SML3403SML3326SML0688
description

≥98% (HPLC)

description

≥95% (HPLC)

description

≥90% (HPLC)

description

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥95% (HPLC)

assay

≥90% (HPLC)

assay

≥98% (HPLC)

form

powder

form

powder

form

powder

form

powder

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

storage temp.

2-8°C

storage temp.

-10 to -25°C

storage temp.

2-8°C

storage temp.

2-8°C

storage condition

protect from light

storage condition

protect from light

storage condition

-

storage condition

-

solubility

DMSO: 2 mg/mL, clear

solubility

H2O: 2 mg/mL, clear

solubility

DMSO: 2 mg/mL, clear (Warmed)

solubility

DMSO: 1 mg/mL, clear (warmed)


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters