Skip to Content
Merck
All Photos(1)

Key Documents

SML3336

Sigma-Aldrich

VU0155056

≥98% (HPLC)

Synonym(s):

5W0, ML272, N-(2-{4-[2-Oxo-2,3-dihydro-1H-benzo(d)imidazol-1-yl]piperidin-1-yl}ethyl)-2-naphthamide, N-[2-[4-(2,3-Dihydro-2-oxo-1H-benzimidazol-1-yl)-1-piperidinyl]ethyl]-2-naphthalenecarboxamide, VU 0155056

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H26N4O2
CAS Number:
Molecular Weight:
414.50
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

Biochem/physiol Actions

VU0155056 is a catalytic site-targeting phospholipase D inhibitor (PLD1/PLD2 IC50 = 81 nM/240 nM by cell-free enzymatic assays; substrate = di-palmitoyl-PLC) that effectively suppresses the cellular PLD activity in GFP-PLD2-overexpressing HEK293 cells as well as PMA-stimulated non-small-cell lung cancer (NSCLC) Calu-1 cells with predominant PLD1 activity (IC50 = 380 nM and 21 nM, respectively).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

No data available

Flash Point(C)

No data available


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jeffrey L Barr et al.
International journal of molecular sciences, 22(10) (2021-06-03)
Orexin A, an endogenous peptide involved in several functions including reward, acts via activation of orexin receptors OX1 and OX2, Gq-coupled GPCRs. We examined the effect of a selective OX1 agonist, OXA (17-33) on cytosolic calcium concentration, [Ca2+]i, in neurons
Priyanka Rai et al.
Proceedings of the National Academy of Sciences of the United States of America, 114(49), 12958-12963 (2017-11-22)
Despite massive fluctuations in its internal triglyceride content, the liver secretes triglyceride under tight homeostatic control. This buffering function is most visible after fasting, when liver triglyceride increases manyfold but circulating serum triglyceride barely fluctuates. How the liver controls triglyceride
Najim Lahrouchi et al.
The Journal of clinical investigation, 131(5) (2021-03-02)
Congenital heart disease is the most common type of birth defect, accounting for one-third of all congenital anomalies. Using whole-exome sequencing of 2718 patients with congenital heart disease and a search in GeneMatcher, we identified 30 patients from 21 unrelated
Amanda R Noble et al.
British journal of cancer, 118(2), 189-199 (2017-11-15)
Phospholipases D1 and D2 (PLD1/2) hydrolyse cell membrane glycerophospholipids to generate phosphatidic acid, a signalling lipid, which regulates cell growth and cancer progression through effects on mTOR and PKB/Akt. PLD expression and/or activity is raised in breast, colorectal, gastric, kidney
Mukesh Kumar et al.
The Journal of cell biology, 218(11), 3697-3713 (2019-10-13)
Triglyceride-rich lipid droplets (LDs) are catabolized with high efficiency in hepatocytes to supply fatty acids for producing lipoprotein particles. Fasting causes a massive influx of adipose-derived fatty acids into the liver. The liver in the fasted state is therefore bloated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service