Skip to Content
Merck
All Photos(1)

Key Documents

SML3251

Sigma-Aldrich

GRL-1720

≥98% (HPLC)

Synonym(s):

1H-Indole-4-carboxylic acid 5-chloro-3-pyridinyl ester, 5-Chloro-3-pyridinyl 1H-indole-4-carboxylate, GRL 0496, GRL-0496, GRL0496

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H9ClN2O2
CAS Number:
Molecular Weight:
272.69
MDL number:
UNSPSC Code:
51111800
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: ≥2 mg/mL, clear

storage temp.

2-8°C

SMILES string

ClC1=CN=CC(OC(C2=CC=CC3=C2C=CN3)=O)=C1

InChI key

BOSZJNSICHFHMA-UHFFFAOYSA-N

Biochem/physiol Actions

GRL-1720 is an irreversible, covalent inhibitor of SARS-CoV-2 Mpro that blocks virus replication. GRL-1720 protects VeroE6 cells form SARS-CoV-2WK-521 infection. It shows no significant cytotoxicity to several cell lines.
Irreversible, covalent inhibitor of SARS-CoV-2 Mpro that blocks virus replication

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Arun K Ghosh et al.
Bioorganic & medicinal chemistry letters, 18(20), 5684-5688 (2008-09-18)
Design, synthesis and biological evaluation of a series of 5-chloropyridine ester-derived severe acute respiratory syndrome-coronavirus chymotrypsin-like protease inhibitors is described. Position of the carboxylate functionality is critical to potency. Inhibitor 10 with a 5-chloropyridinyl ester at position 4 of the
Marina Macchiagodena et al.
Journal of chemical theory and computation, 16(11), 7160-7172 (2020-10-23)
In the context of drug-receptor binding affinity calculations using molecular dynamics techniques, we implemented a combination of Hamiltonian replica exchange (HREM) and a novel nonequilibrium alchemical methodology, called virtual double-system single-box, with increased accuracy, precision, and efficiency with respect to
Shin-Ichiro Hattori et al.
Nature communications, 12(1), 668-668 (2021-01-30)
Except remdesivir, no specific antivirals for SARS-CoV-2 infection are currently available. Here, we characterize two small-molecule-compounds, named GRL-1720 and 5h, containing an indoline and indole moiety, respectively, which target the SARS-CoV-2 main protease (Mpro). We use VeroE6 cell-based assays with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service