Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|---|---|
5 mg | Check Cart for Availability | € 86,60 |
25 mg | Check Cart for Availability | € 349,00 |
About This Item
Empirical Formula (Hill Notation):
C24H23N3O3
Molecular Weight:
401.46
UNSPSC Code:
12352107
NACRES:
NA.77
Assay:
≥98% (HPLC)
Form:
powder
Quality Segment
assay
≥98% (HPLC)
form
powder
color
white to beige
solubility
DMSO: 2 mg/mL, clear
storage temp.
2-8°C
SMILES string
OCCCC#CC1=CC=CC=C1C2=NN3C(COC(C=C(NC(C)=O)C=C4)=C4C3)=C2
InChI
1S/C24H23N3O3/c1-17(29)25-20-11-10-19-15-27-21(16-30-24(19)13-20)14-23(26-27)22-9-5-4-8-18(22)7-3-2-6-12-28/h4-5,8-11,13-14,28H,2,6,12,15-16H2,1H3,(H,25,29)
InChI key
VVMOXIHBNMUNFJ-UHFFFAOYSA-N
Biochem/physiol Actions
NA-11 is a potent and selective activator of PFKL (phosphofructokinase-1 liver type) that suppress excessive NOX2-dependent oxidative burst and neutrophil cell death (NETosis). NA-11 decreases neutrophiles efficiency to kill bacteria. It prevents neutrophil-induced tissue damage.
potent and selective activator of PFKL (phosphofructokinase-1 liver type) that suppress excessive NOX2-dependent oxidative burst
1 of 1
This Item | |||
|---|---|---|---|
| assay ≥98% (HPLC) | assay ≥98% (HPLC) | assay ≥98% (HPLC) | assay ≥98% (HPLC) |
| form powder | form powder | form powder | form powder |
| Quality Level 100 | Quality Level 100 | Quality Level 100 | Quality Level 100 |
| storage temp. 2-8°C | storage temp. 2-8°C | storage temp. 2-8°C | storage temp. 2-8°C |
| solubility DMSO: 2 mg/mL, clear | solubility DMSO: 2 mg/mL, clear | solubility DMSO: 2 mg/mL, clear | solubility DMSO: 1 mg/mL, clear (warmed) |
| color white to beige | color white to beige | color white to beige | color white to beige |
Storage Class
11 - Combustible Solids
wgk
WGK 3
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.



