Skip to Content
Merck

Skip To

SML3232

BAY-6672 hydrochloride hydrate

≥98% (HPLC)

Synonym(s):

(+)-(4R)-5-({[6-Bromo-3-methyl-2-(pyrrolidin-1-yl)quinolin-4-yl]carbonyl}amino)-4-(2-chlorophenyl)pentanoic acid, BAY 6672 hydrochloride hydrate, BAY6672 hydrochloride hydrate, hydrochloride hydrate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
5 mg
Check Cart for Availability
€ 105,00
25 mg
Check Cart for Availability
€ 393,00

About This Item

Empirical Formula (Hill Notation):
C26H27BrClN3O3 · HCl · H2O
CAS Number:
Molecular Weight:
599.34
UNSPSC Code:
12352107
NACRES:
NA.77
Assay:
≥98% (HPLC)
Form:
powder
Storage condition:
desiccated

€ 105,00


Check Cart for Availability

Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

ClC1=CC=CC=C1[C@@H](CCC(O)=O)CNC(C2=C(C)C(N3CCCC3)=NC4=CC=C(Br)C=C42)=O.Cl.O

InChI

1S/C26H27BrClN3O3.ClH.H2O/c1-16-24(20-14-18(27)9-10-22(20)30-25(16)31-12-4-5-13-31)26(34)29-15-17(8-11-23(32)33)19-6-2-3-7-21(19)28;;/h2-3,6-7,9-10,14,17H,4-5,8,11-13,15H2,1H3,(H,29,34)(H,32,33);1H;1H2/t17-;;/m0../s1

InChI key

GGGPATBQZZPKRC-RMRYJAPISA-N

Biochem/physiol Actions

BAY-6672 is an orally active, potent and selective prostaglandin F receptor (FP-R, PTGFR) antagonist (IC50 = 11 nM against 3 nM PGF2?-induced calcium flux in hFP-R Chem-1 cells; IC50 = 12/18 nM against 3 μM PGF2α-induced KC/MCP1 production in NIH-3T3) with >420-fold selectivity over human prostaglandin receptors (PG-Rs) EP1-4, DP, CRTH2 and IP. BAY-6672 shows in vivo efficacy in a murine lung fibrosis model (3, 10, 30 mg/kg b.i.d. p.o.).
Orally active, potent and selective prostaglandin F receptor (FP-R, PTGFR) antagonist with in vivo efficacy in a murine lung fibrosis model.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
S4696G5793A9736
assay

≥98% (HPLC)

assay

≥98% (HPLC)

assay

>98% (HPLC)

assay

≥98% (HPLC)

form

powder

form

powder

form

powder

form

powder

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage condition

desiccated

storage condition

desiccated

storage condition

desiccated

storage condition

desiccated

solubility

DMSO: 2 mg/mL, clear

solubility

DMSO: >10 mg/mL, H2O: insoluble

solubility

DMSO: 10 mg/mL, clear

solubility

DMSO: 20 mg/mL, clear


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters