Skip to Content
Merck
All Photos(1)

Key Documents

SML3212

Sigma-Aldrich

Eliglustat

≥98% (HPLC)

Synonym(s):

Genz 99067, N-[(1R,2R)-2-(2,3-Dihydro-1,4-benzodioxin-6-yl)-2-hydroxy-1-(1-pyrrolidinylmethyl)ethyl]-octanamide

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€78.50
25 MG
€253.00

€78.50


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
5 MG
€78.50
25 MG
€253.00

About This Item

Empirical Formula (Hill Notation):
C23H36N2O4
CAS Number:
Molecular Weight:
404.54
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

€78.50


Estimated to ship on28 May 2025


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

N3(CCCC3)C[C@@H](NC(=O)CCCCCCC)[C@H](O)c1cc2c(cc1)OCCO2

InChI

1S/C23H36N2O4/c1-2-3-4-5-6-9-22(26)24-19(17-25-12-7-8-13-25)23(27)18-10-11-20-21(16-18)29-15-14-28-20/h10-11,16,19,23,27H,2-9,12-15,17H2,1H3,(H,24,26)/t19-,23-/m1/s1

InChI key

FJZZPCZKBUKGGU-AUSIDOKSSA-N

Biochem/physiol Actions

Eliglustat is a potent and selctive glucosylceramide synthase inhibitor approved for treatment of type 1 Gaucher disease.
Glucosylceramide synthase inhibitor, used for treatment of type 1 Gaucher disease

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - STOT RE 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service