Skip to Content
Merck
All Photos(1)

Key Documents

SML3208

Sigma-Aldrich

CPYPP

≥98% (HPLC)

Synonym(s):

4-[3′-(2′′-chlorophenyl)-2′-propen-1′-ylidene]-1-phenyl-3,5-pyrazolidinedione

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€78.60
25 MG
€317.00

€78.60


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
€78.60
25 MG
€317.00

About This Item

Empirical Formula (Hill Notation):
C18H13ClN2O2
CAS Number:
Molecular Weight:
324.76
MDL number:
UNSPSC Code:
51111800
NACRES:
NA.77

€78.60


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

orange to dark red

solubility

DMSO: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

Clc1c(cccc1)C=CC=C2C(=O)N(NC2=O)c3ccccc3

InChI

1S/C18H13ClN2O2/c19-16-12-5-4-7-13(16)8-6-11-15-17(22)20-21(18(15)23)14-9-2-1-3-10-14/h1-12H,(H,20,22)

InChI key

VVZJFICTTKPNCK-UHFFFAOYSA-N

Biochem/physiol Actions

CPYPP is a cell permeable potent and selective reversible inhibitor of DOCK2 that binds to DOCK2 DHR-2 domain. CPYPP is a DOCK2-Rac1 interaction inhibitor. It also inhibits DOCK1 and DOCK5. CPYPP potently inhibits metastatic spread of aggressive ROBO1deficient prostate cancer cells through the vasculature of transgenic Zebrafish model.
cell permeable potent and selective reversible inhibitor of DOCK2 that binds to DOCK2 DHR-2 domain

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service