Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|---|---|
5 mg | Check Cart for Availability | € 81,60 |
25 mg | Check Cart for Availability | € 329,00 |
About This Item
Empirical Formula (Hill Notation):
C18H13F2NO3
CAS Number:
Molecular Weight:
329.30
UNSPSC Code:
12352106
NACRES:
NA.77
MDL number:
Assay:
≥98% (HPLC)
Form:
powder
Quality Segment
assay
≥98% (HPLC)
form
powder
color
white to beige
solubility
DMSO: 2 mg/mL, clear
storage temp.
2-8°C
SMILES string
OC(C(CC(C1=C(F)C=C(F)C=C1)=O)C2=CNC3=CC=CC=C32)=O
InChI
1S/C18H13F2NO3/c19-10-5-6-12(15(20)7-10)17(22)8-13(18(23)24)14-9-21-16-4-2-1-3-11(14)16/h1-7,9,13,21H,8H2,(H,23,24)
InChI key
BOKQALWNGNLTOC-UHFFFAOYSA-N
Biochem/physiol Actions
Mitochonic Acid 5 is a potent enhancer of mitochondrial ATP production that significantly increased the cellular ATP level in Hep3B human hepatocellular carcinoma cells. Mitochonic Acid 5 improves the survival of fibroblasts from patients with mitochondrial diseases. It appears that mitochonic Acid 5 facilitates ATP synthase oligomerization and supercomplex formation with mitofilin/Mic60.
potent enhancer of mitochondrial ATP production
1 of 1
This Item | |||
|---|---|---|---|
| description ≥98% (HPLC) | description ≥98% (HPLC) | description ≥98% (HPLC) | description ≥97% (HPLC) |
| assay ≥98% (HPLC) | assay ≥98% (HPLC) | assay ≥98% (HPLC) | assay ≥97% (HPLC) |
| form powder | form powder | form powder | form powder |
| Quality Level 100 | Quality Level 100 | Quality Level 100 | Quality Level 100 |
| storage temp. 2-8°C | storage temp. 2-8°C | storage temp. 2-8°C | storage temp. 2-8°C |
| solubility DMSO: 2 mg/mL, clear | solubility DMSO: 2 mg/mL, clear | solubility DMSO: 2 mg/mL, clear | solubility DMSO: 2 mg/mL, clear |
| color white to beige | color white to beige | color white to beige | color white to beige |
Storage Class
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.



