Skip to Content
Merck
All Photos(1)

Key Documents

SML3127

Sigma-Aldrich

Barbadin

≥98% (HPLC)

Synonym(s):

3-Amino-5-(4-benzylphenyl)thieno[2,3-d]pyrimidin-4-one, 3-Amino-5-[4-(phenylmethyl)phenyl]thieno[2,3-d]pyrimidin-4(3H)-one

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€88.40
25 MG
€357.00

€88.40


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
€88.40
25 MG
€357.00

About This Item

Empirical Formula (Hill Notation):
C19H15N3OS
CAS Number:
Molecular Weight:
333.41
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

€88.40


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C1N(C=NC2=C1C(C3=CC=C(C=C3)CC4=CC=CC=C4)=CS2)N

InChI

1S/C19H15N3OS/c20-22-12-21-18-17(19(22)23)16(11-24-18)15-8-6-14(7-9-15)10-13-4-2-1-3-5-13/h1-9,11-12H,10,20H2

InChI key

OCBXPCSXEQQADU-UHFFFAOYSA-N

Biochem/physiol Actions

selective b-arrestin/b2-adaptin inhibitor

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service