Skip to Content
Merck
All Photos(1)

Key Documents

SML3120

Sigma-Aldrich

LY272015

≥98% (HPLC)

Synonym(s):

1-[(3,4-Dimethoxyphenyl)methyl]-2,3,4,9-tetrahydro-6-methyl-1H-pyrido[3,4-b]indole monohydrochloride, 1H-Pyrido[3,4-b]indole, 1-[(3,4-dimethoxyphenyl)methyl]-2,3,4,9-tetrahydro-6-methyl-, hydrochloride (1:1), LY 272015, LY-272015

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€86.60
25 MG
€349.00

€86.60


Estimated to ship on23 May 2025


Request a Bulk Order

Select a Size

Change View
5 MG
€86.60
25 MG
€349.00

About This Item

Empirical Formula (Hill Notation):
C21H24N2O2 · HCl
CAS Number:
Molecular Weight:
372.89
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

€86.60


Estimated to ship on23 May 2025


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

CC1=CC(C2=C(C(NCC2)CC(C=C3OC)=CC=C3OC)N4)=C4C=C1.Cl

InChI

1S/C21H24N2O2.ClH/c1-13-4-6-17-16(10-13)15-8-9-22-18(21(15)23-17)11-14-5-7-19(24-2)20(12-14)25-3;/h4-7,10,12,18,22-23H,8-9,11H2,1-3H3;1H

InChI key

BKAZOTIBKRWLQA-UHFFFAOYSA-N

Biochem/physiol Actions

High-affinity, 5-HT2B subtype-selective serotonin receptor antagonist with in vitro and in vivo efficacy.
LY272015 is a high-affinity, 5-HT2B subtype-selective serotonin receptor antagonist (5-HT2B -log KB = 9.86, 5-HT2A/5-HT2C -log Ki = 7.65/7.92) that exhibits antihypertensive efficacy in deoxycorticosterone acetate (DOCA)-salt-hypertensive rats in vivo (1, 3 mg/kg iv; 1x/wk for 4 wks). LY272015 inhibits mitral valve interstitial cell (MVIC) activation by 5HT in cultures (100 nM LY, 10 nM or 1 μM 5HT; canine MVIC) and ex vivo (100 μM LY, 10 μM 5HT; human MV), as well as mitigates angiotensin II infusion-provoked MV remodeling in mice in vivo (3 mg/kg via infusion by osmotic pumps 2x/wk).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service