Skip to Content
Merck
All Photos(1)

Key Documents

SML3106

Sigma-Aldrich

7ACC2

≥98% (HPLC)

Synonym(s):

7-(N-Benzyl-N-methylamino)-2-oxo-2H-chromene-3-carboxylic acid, 7-ACC2, 7-Aminocarboxycoumarin 2, 7-[Benzyl(methyl)amino]-2-oxo-2H-chromene-3-carboxylic acid, 7-[Methyl(phenylmethyl)amino]-2-oxo-2H-1-benzopyran-3-carboxylic acid

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€78.50
25 MG
€253.00

€78.50


Estimated to ship on12 April 2025


Request a Bulk Order

Select a Size

Change View
5 MG
€78.50
25 MG
€253.00

About This Item

Empirical Formula (Hill Notation):
C18H15NO4
CAS Number:
Molecular Weight:
309.32
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

€78.50


Estimated to ship on12 April 2025


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C(O)C1=CC2=C(C=C(C=C2)N(C)CC3=CC=CC=C3)OC1=O

InChI

1S/C18H15NO4/c1-19(11-12-5-3-2-4-6-12)14-8-7-13-9-15(17(20)21)18(22)23-16(13)10-14/h2-10H,11H2,1H3,(H,20,21)

InChI key

XTKDQPFUOFAMRL-UHFFFAOYSA-N

Biochem/physiol Actions

7ACC2 is a monocarboxylate transporters MCT1 & MCT4 (SLC16A1 & SLC16A3) inhibitor that affects lactate uptake-dependent cancer growth in cultures (uptake/proliferation IC50 = 59/220 nM; MCT1/4-expressing SiHA cervix cancer in the absence of glucose) and in vivo (3 mg/kg/day i.p.; SiHa, HCT116, MCF-7 in mice), without affecting lactate efflux or the survival of highly glycolytic HL-60 (MCT1) or MDA-MB231 (MCT4) cancer cells. Unlike the MCT1/2 inhibitor AR-C155858, 7ACC2 does not block MCT1-mediated uptake of the anti-cancer drug 3-bromopyruvate (3-BP).
Monocarboxylate transporters MCT1 & MCT4 (SLC16A1 & SLC16A3) inhibitor that blocks lactate update-dependent cancer growth in vitro and in vivo.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service