Skip to Content
Merck
All Photos(1)

Key Documents

SML3098

Sigma-Aldrich

C-178

≥98% (HPLC)

Synonym(s):

C 178, C178, N-(Dibenzo[b,d]furan-3-yl)-5-nitrofuran-2-carboxamide, N-3-Dibenzofuranyl-5-nitro-2-furancarboxamide, N-Dibenzofuran-3-yl-5-nitrofuran-2-carboxamide

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€78.40
25 MG
€253.00

€78.40


Estimated to ship on16 April 2025


Request a Bulk Order

Select a Size

Change View
5 MG
€78.40
25 MG
€253.00

About This Item

Empirical Formula (Hill Notation):
C17H10N2O5
CAS Number:
Molecular Weight:
322.27
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

€78.40


Estimated to ship on16 April 2025


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

faint yellow to dark brown

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C(C1=CC=C([N+]([O-])=O)O1)NC2=CC(OC3=C4C=CC=C3)=C4C=C2

InChI key

URUVDCCYSJEGQQ-UHFFFAOYSA-N

Biochem/physiol Actions

C-178 is a potent and selective inhibitor against murine, but not human, stimulator of interferon genes (STING) via covalent modifiation of mSTING Cys91. C-178 inhibits mSTING-mediated IFNβ reporter activity (IC50 <500 nM) without affecting RIG-I, and blocks (0.5 μM) downstream TBK1 phosphorylation induction by STING agonist CMA (1.4 h) in mSTING-expressing HEK293T cells.
Potent and selective, Cys91-targeting covalent inhibitor against murine, but not human, STING.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yajuan Fu et al.
iScience, 23(4), 101026-101026 (2020-04-14)
DNA transfection is often the bottleneck of research and gene therapy practices. To explore the mechanism regulating transgene expression, we investigated the role of the cGAS-STING signaling pathway, which induces type-I interferons in response to DNA. We confirmed that deletion
Simone M Haag et al.
Nature, 559(7713), 269-273 (2018-07-06)
Aberrant activation of innate immune pathways is associated with a variety of diseases. Progress in understanding the molecular mechanisms of innate immune pathways has led to the promise of targeted therapeutic approaches, but the development of drugs that act specifically
Giulia Maria Piperno et al.
JCI insight, 5(17) (2020-07-30)
Dysregulated sensing of self-nucleic acid is a leading cause of autoimmunity in multifactorial and monogenic diseases. Mutations in Wiskott-Aldrich syndrome protein (WASp), a key regulator of cytoskeletal dynamics in immune cells, cause autoimmune manifestations and increased production of type I
Kojiro Mukai et al.
Nature communications, 12(1), 61-61 (2021-01-06)
Coat protein complex I (COP-I) mediates the retrograde transport from the Golgi apparatus to the endoplasmic reticulum (ER). Mutation of the COPA gene, encoding one of the COP-I subunits (α-COP), causes an immune dysregulatory disease known as COPA syndrome. The

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service