Skip to Content
Merck
All Photos(1)

Key Documents

SML3092

Sigma-Aldrich

CHIR-090

≥98% (HPLC)

Synonym(s):

CHIR 090, CHIR090, N-((2S,3R)-3-Hydroxy-1-(hydroxyamino)-1-oxobutan-2-yl)-4-((4-(morpholinomethyl)phenyl)ethynyl)benzamide, N-[(1S,2R)-2-Hydroxy-1-[(hydroxyamino)carbonyl]propyl]-4-[2-[4-(4-morpholinylmethyl)phenyl]ethynyl]benzamide

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€128.00
25 MG
€516.00

€128.00


Estimated to ship on17 May 2025


Request a Bulk Order

Select a Size

Change View
5 MG
€128.00
25 MG
€516.00

About This Item

Empirical Formula (Hill Notation):
C24H27N3O5
CAS Number:
Molecular Weight:
437.49
MDL number:
UNSPSC Code:
51111800
NACRES:
NA.77

€128.00


Estimated to ship on17 May 2025


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

C[C@H]([C@@H](C(NO)=O)NC(C1=CC=C(C=C1)C#CC2=CC=C(C=C2)CN3CCOCC3)=O)O

InChI

1S/C24H27N3O5/c1-17(28)22(24(30)26-31)25-23(29)21-10-8-19(9-11-21)3-2-18-4-6-20(7-5-18)16-27-12-14-32-15-13-27/h4-11,17,22,28,31H,12-16H2,1H3,(H,25,29)(H,26,30)/t17-,22+/m1/s1

InChI key

FQYBTYFKOHPWQT-VGSWGCGISA-N

Biochem/physiol Actions

CHIR-090 is a potent, tight-binding inhibitor against LpxC, the zinc-dependent UDP-3-O-(R-3-hydroxymyristoyl)-N-acetylglucosamine deacetylase essential for gram negative bacteria lipid A biosynthesis (98% inhibition of E. coli & A. aeolicus LpxC by 0.5 μg/mL (1.15 μM) CHIR-090 at pH 7.4; A. aeolicus LpxC Ki = 1.0-1.7 nM). CHIR-090 possesses remarkable antibiotic activity comparable to that of ciprofloxacin against P. aeruginosa PAO1 as well as E. coli R477 and R477 [LpxC G17S] strains by antibacterial disk tests (20 μg/2 μL per spot).
Potent, tight-binding LpxC inhibitor with remarkable antibiotic activity comparable to that of ciprofloxacin.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service