Skip to Content
Merck
All Photos(1)

Key Documents

SML3085

Sigma-Aldrich

Evacetrapib

≥95% (HPLC)

Synonym(s):

LY 2484595, LY-2484595, LY2484595, trans-4-({(5S)-5-[{[3,5-bis(Trifluoromethyl)phenyl]methyl}(2-methyl-2H-tetrazol-5-yl)amino]-7,9-dimethyl-2,3,4,5-tetrahydro-1H-benzazepin-1-yl}methyl)cyclohexanecarboxylic acid, trans-4-[[(5S)-5-[[[3,5-bis(Trifluoromethyl)phenyl]methyl](2-methyl-2H-tetrazol-5-yl)amino]-2,3,4,5-tetrahydro-7,9-dimethyl-1H-1-benzazepin-1-yl]methyl]-cyclohexanecarboxylic acid

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
€78.70
50 MG
€317.00

€78.70


Estimated to ship on21 March 2025


Request a Bulk Order

Select a Size

Change View
10 MG
€78.70
50 MG
€317.00

About This Item

Empirical Formula (Hill Notation):
C31H36F6N6O2
CAS Number:
Molecular Weight:
638.65
MDL number:
UNSPSC Code:
51111800
NACRES:
NA.77

€78.70


Estimated to ship on21 March 2025


Request a Bulk Order

Quality Level

Assay

≥95% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

CC1=C2C([C@@H](N(CC3=CC(C(F)(F)F)=CC(C(F)(F)F)=C3)C4=NN(C)N=N4)CCCN2C[C@H]5CC[C@H](C(O)=O)CC5)=CC(C)=C1

InChI

1S/C31H36F6N6O2/c1-18-11-19(2)27-25(12-18)26(5-4-10-42(27)16-20-6-8-22(9-7-20)28(44)45)43(29-38-40-41(3)39-29)17-21-13-23(30(32,33)34)15-24(14-21)31(35,36)37/h11-15,20,22,26H,4-10,16-17H2,1-3H3,(H,44,45)/t20?,22?,26-/m0/s1

InChI key

IHIUGIVXARLYHP-UXNJHFGPSA-N

Biochem/physiol Actions

Evacetrapib (LY2484595) is an orally active, potent and selective cholesteryl ester transfer protein (CETP) inhibitor (human recombinant CETP IC50 = 5.5 nM). Evacetrapib inhibits CETP activtiy in human plasma ex vivo (IC50 = 36 nM) and in human CETP/ApoAI transgenic mice in vivo (129.7% HDL-C increase 8 h post oral dosing at 30 mg/kg; 98.4%, 98.6%, 18.4% CETP inhibition, respectivley, post 4, 8 and 24 h p.o.). Evacetrapib does not cause the side-effects observed with torcetrapib in cultures (no aldosterone or cortisol synthesis induction in H295R cells) or in vivo (no increase of mean arterial pressure in diabetic fatty rats).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service