Skip to Content
Merck
All Photos(1)

Key Documents

SML3068

Sigma-Aldrich

PSEM308 hydrochloride

≥98% (HPLC)

Synonym(s):

5-Methyl-3,4,14-triazapentacyclo[12.2.2.14,7.02,13.011,19]nonadeca-2,7(19),8,10-tetraene hydrochloride, 5-Methyl-5,8,9,10,11a,12-hexahydro-4H-8,11-ethanopyrido[3′,2′:3,4]diazepino[6,7,1-hi]indole hydrochloride, PSEM 308 hydrochloride, PSEM308 hydrochloride, Pharmacologically selective effector molecule 308, hydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€102.00
25 MG
€413.00

€102.00


Estimated to ship on23 May 2025


Request a Bulk Order

Select a Size

Change View
5 MG
€102.00
25 MG
€413.00

About This Item

Empirical Formula (Hill Notation):
C17H21N3 · HCl
Molecular Weight:
303.83
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

€102.00


Estimated to ship on23 May 2025


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

DMSO: 2 mg/mL, clear (warmed)

storage temp.

2-8°C

SMILES string

CC1CC2=CC=CC3=C2N1N=C(C4CCN5CC4)C5C3.Cl

Biochem/physiol Actions

PSEM agonist for chimeric ligand-gated ion channel pores with nAChR ?7 LBD L141F or L141F/Y115F mutant (PSAM) in vitro and in vivo.
SEM308 is a pharmacologically selective effector molecule (PSEM) that acts as a selective agonist in vitro and in vivo for chimeric pharmacologically selective actuator modules (PSAMs) composed of nAChR α7 ligand-binding domain (LBD) with L141F or L141F/Y115F mutation fused to the ion pore domain (IPD) of a ligand-gated ion channel (LGIC), including glycine receptor (PSAML141F-GlyR & PSAML141F/Y115F-GlyR) and 5-HT3 (PSAML141F/Y115F-5-HT3).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Maria Soledad Esposito et al.
Nature, 508(7496), 351-356 (2014-02-04)
Translating the behavioural output of the nervous system into movement involves interaction between brain and spinal cord. The brainstem provides an essential bridge between the two structures, but circuit-level organization and function of this intermediary system remain poorly understood. Here
Kajana Satkunendrarajah et al.
Nature, 562(7727), 419-422 (2018-10-12)
Dysfunctional breathing is the main cause of morbidity and mortality after traumatic injury of the cervical spinal cord1,2 and often necessitates assisted ventilation, thus stressing the need to develop strategies to restore breathing. Cervical interneurons that form synapses on phrenic
Daisuke Satoh et al.
Neuron, 89(5), 1046-1058 (2016-03-01)
The most commonly used locomotor strategy in rodents is left-right limb alternation. Mutation of the axon guidance molecule EphA4 profoundly alters this basic locomotor pattern to synchrony. Here we report that conditional mutation of EphA4 in spinal interneurons expressing the
Akila Chandrasekar et al.
Cerebral cortex (New York, N.Y. : 1991), 29(6), 2701-2715 (2018-07-10)
Excessive excitation has been hypothesized to subsume a significant part of the acute damage occurring after traumatic brain injury (TBI). However, reduced neuronal excitability, loss of neuronal firing, and a disturbed excitation/inhibition balance have been detected. Parvalbumin (PV) interneurons are
Matthew Lovett-Barron et al.
Nature neuroscience, 15(3), 423-430 (2012-01-17)
Transforming synaptic input into action potential output is a fundamental function of neurons. The pattern of action potential output from principal cells of the mammalian hippocampus encodes spatial and nonspatial information, but the cellular and circuit mechanisms by which neurons

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service