Skip to Content
Merck
All Photos(1)

Key Documents

SML3009

Sigma-Aldrich

Indiplon

≥98% (HPLC)

Synonym(s):

CL 285,489, CL 285489, CL-285,489, CL-285489, CL285,489, CL285489, N-Methyl-N-(3-(3-(thiophene-2-carbonyl)pyrazolo[1,5-a]pyrimidin-7-yl)phenyl)acetamide, N-Methyl-N-[3-[3-(2-thienylcarbonyl)pyrazolo[1,5-a]pyrimidin-7-yl]phenyl]acetamide, NBI 34060, NBI-34060, NBI34060

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€70.60
25 MG
€229.00

€70.60


Estimated to ship on26 April 2025


Request a Bulk Order

Select a Size

Change View
5 MG
€70.60
25 MG
€229.00

About This Item

Empirical Formula (Hill Notation):
C20H16N4O2S
CAS Number:
Molecular Weight:
376.43
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

€70.60


Estimated to ship on26 April 2025


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

[s]1c(ccc1)C(=O)c2c3[n](nc2)C(=CC=N3)c4cc(ccc4)N(C)C(=O)C

InChI

1S/C20H16N4O2S/c1-13(25)23(2)15-6-3-5-14(11-15)17-8-9-21-20-16(12-22-24(17)20)19(26)18-7-4-10-27-18/h3-12H,1-2H3

InChI key

CBIAWPMZSFFRGN-UHFFFAOYSA-N

Biochem/physiol Actions

Indiplon (NBI 34060) is an orally active, high-affinity GABAA receptor positive allosteric modulator (PAM) with preferential labeling of alpha1 subunits-containing receptors (rat cerebellar/cerebral cortex membranes binding KD = 1.01/0.45 nM). Indiplon potentiates GABA-induced chloride conductance of cultured rat cortical neurons (EC50 = 11.6 nM vs 152 nM/630 nM) and exhibits sedative efficacy in mice (ED50 = 1 mg/kg/passive avoidance & 2.7 mg/kg/locomotor activity inhibition po.) and rats (ED50 = 2.5 mg/kg/locomotor activity inhibition & 3 mg/kg/vigilance impairment po.) in vivo. Indiplon shows greater affinity, in vitor and in vivo potency than zaleplon and zolpidem.
Orally active, high-affinity GABA(A) receptor positive allosteric modulator (PAM) with in vivo sedative-hypnotic efficacy.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alan C Foster et al.
The Journal of pharmacology and experimental therapeutics, 311(2), 547-559 (2004-07-17)
Indiplon (NBI 34060; N-methyl-N-[3-[3-(2-thienylcarbonyl)-pyrazolo[1,5-alpha]pyrimidin-7-yl]phenyl]acetamide), a novel pyrazolopyrimidine and high-affinity allosteric potentiator of GABA(A) receptor function, was profiled for its effects in rodents after oral administration. In mice, indiplon inhibited locomotor activity (ED(50) = 2.7 mg/kg p.o.) at doses lower than
D K Cass et al.
Molecular psychiatry, 19(5), 536-543 (2014-03-05)
Converging epidemiological studies indicate that cannabis abuse during adolescence increases the risk of developing psychosis and prefrontal cortex (PFC)-dependent cognitive impairments later in life. However, the mechanisms underlying the adolescent susceptibility to chronic cannabis exposure are poorly understood. Given that
Ignacio Vega-Quiroga et al.
Neuropharmacology, 128, 76-85 (2017-10-01)
The mechanisms commanding the activity of dopaminergic neurons of the ventral tegmental area (VTA) and the location of these neurons are relevant for the coding and expression of motivated behavior associated to reward-related signals. Anatomical evidence shows that several brain
Robert E Petroski et al.
The Journal of pharmacology and experimental therapeutics, 317(1), 369-377 (2006-01-10)
Indiplon (NBI 34060) is a novel pyrazolopyrimidine currently in development for the treatment of insomnia. We have previously shown that indiplon exhibits high-affinity binding to native GABA(A) receptors from rat brain and acts as a positive allosteric modulator of GABA(A)
Yanbo Jiang et al.
Neuropharmacology, 138, 97-105 (2018-06-09)
Ionotropic GABAA receptors expressing at the axon initial segment (AIS) of glutamatergic pyramidal cell (PC) in the cortex plays critical roles in regulating action potential generation. However, it remains unclear whether these receptors also express at the AIS of cortical

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service