Skip to Content
Merck

Skip To

SML2963

8-Azaguanine

≥98% (HPLC)

Synonym(s):

5-Amino-1H-v-triazolo[d]pyrimidin-7-ol, 5-Amino-3,6-dihydro-7H-1,2,3-triazolo[4,5-d]pyrimidin-7-one, 5-Amino-7-hydroxy-1H-v-triazolo[d]pyrimidine, 8-AG, Guanazolo, NSC 151069, NSC 223526, pathocidin, 2-Amino-6-hydroxy-8-azapurine, 2-Amino-6-oxy-8-azapurine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
10 mg
Check Cart for Availability
€ 42,00
50 mg
Check Cart for Availability
€ 171,00

About This Item

Empirical Formula (Hill Notation):
C4H4N6O
CAS Number:
Molecular Weight:
152.11
UNSPSC Code:
12352200
Beilstein/REAXYS Number:
171098
MDL number:
NACRES:
NA.21
Assay:
≥98% (HPLC)
Form:
powder

€ 42,00


Check Cart for Availability

​
Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

mp

>300 °C (lit.)

solubility

DMSO: 2 mg/mL, clear (warmed)

storage temp.

2-8°C

SMILES string

NC1=Nc2[nH]nnc2C(=O)N1

InChI

1S/C4H4N6O/c5-4-6-2-1(3(11)7-4)8-10-9-2/h(H4,5,6,7,8,9,10,11)

InChI key

LPXQRXLUHJKZIE-UHFFFAOYSA-N

Biochem/physiol Actions

8-Azaguanine is a triazole-antimetabolite that displays varies pharmacological effects including anticancer, antiviral and antifungal. It is easily incorporated into ribonucleic acids including rRNA and tRNA. 8-Azaguanine appears to alter mRNA splicing in SF3B1 mutant cells. 8-Azaguanine novel immunomodulatory drug (IMiD) that significantly increases the cytotoxicity of NK cells.


Pictograms

Exclamation mark

Signal Word

Warning

Hazard Codes

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library




Questions

Reviews

No rating value

Active Filters