Skip to Content
Merck
All Photos(1)

Key Documents

SML2959

Sigma-Aldrich

Carbonyl cyanide 4-(trifluoromethoxy)phenylhydrazone Ready Made Solution

≥98% (HPLC), liquid, mithophagy inducer

Synonym(s):

FCCP, Mesoxalonitrile 4-trifluoromethoxyphenylhydrazone

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
€64.30

€64.30


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
1 ML
€64.30

About This Item

Empirical Formula (Hill Notation):
C10H5F3N4O
Molecular Weight:
254.17
UNSPSC Code:
12352200
NACRES:
NA.77

€64.30


Estimated to ship on28 May 2025


Request a Bulk Order

Product Name

Carbonyl cyanide 4-(trifluoromethoxy)phenylhydrazone Ready Made Solution, ≥98% (HPLC), 2.54 mg/mL (10mM) in DMSO

Quality Level

Assay

≥98% (HPLC)

form

liquid

storage temp.

−20°C

General description

Carbonyl cyanide-4-(trifluoromethoxy)phenylhydrazone (FCCP) is a proton (H+) ionophore and a potent mitochondrial uncoupling agent. This compound turns the mitochondrial membrane permeable to protons, thus dissipating the mitochondrial membrane potential and uncouples oxidative phosphorylation from ATP synthesis. FCCP is a potent tool for assaying mitochondrial respiration and investigating the role of mitochondria in cellular function.1,2FCCP induces mitophagy of SH-SY5Y neuroblastoma cell in a dose dependent manner. Low concentration of FCCP (1μM) leads to a complete loss of mitochondrial membrane potential without triggering mitophagy, whereas at a high concentration (10 μM), cytosol acidification is increased and mitochondrial degradation is attained.3 Additionally, FCCP (20 μM) was able to induce apoptosis in lymphoid (CCRF-CEM) and myeloid (HL60) cells after 24hrs of incubation. 4 FCCP (0.5nM-5 μM) reduces β-amyloid peptide production in K695sw cells without altering the β-amyloid precursor protein maturation or synthesis.5 Partial uncoupling of mitochondrial oxidative phosphorylation induced by FCCP (0.3 -3μM) reduces micromotion and wound healing migration of human mesenchymal stem cells.6 Inhibition of F1F0-ATPase by FCCP (10 μM), reduces the impairment of electromechanical activity in human myocardium.7

Biochem/physiol Actions

Mithophagy inducer, ionophore

Other Notes

FCCP ready made solution is a DMSO based solution at 10mM concentration. It is recommended to further dilute in a suitable media to achieve a working concentration of 1-10 μM.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

188.6 °F

Flash Point(C)

87 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service