Skip to Content
Merck

Skip To

SML2941

Maribavir

≥98% (HPLC)

Synonym(s):

(2S,3S,4R,5S)-2-(5,6-Dichloro-2-(isopropylamino)-1H-benzo[d]imidazol-1-yl)-5-(hydroxymethyl)tetrahydrofuran-3,4-diol, 1263W94, Benzimidavir, VP-41263

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
5 mg
Check Cart for Availability
€ 108,00
25 mg
Check Cart for Availability
€ 436,00

About This Item

Empirical Formula (Hill Notation):
C15H19Cl2N3O4
CAS Number:
Molecular Weight:
376.24
NACRES:
NA.77
UNSPSC Code:
12352200
Assay:
≥98% (HPLC)
Form:
powder

€ 108,00


Check Cart for Availability

​
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

CC(NC1=NC2=CC(Cl)=C(Cl)C=C2N1[C@@H]3[C@@H](O)[C@@H](O)[C@H](CO)O3)C

InChI

1S/C15H19Cl2N3O4/c1-6(2)18-15-19-9-3-7(16)8(17)4-10(9)20(15)14-13(23)12(22)11(5-21)24-14/h3-4,6,11-14,21-23H,5H2,1-2H3,(H,18,19)/t11-,12-,13-,14-/m0/s1

InChI key

KJFBVJALEQWJBS-XUXIUFHCSA-N

Biochem/physiol Actions

Maribavir is an orally available antiviral agent under development for treatment and prevention of human cytomegalovirus (HCMV) infection. Maribavir is a potent and selective ATP-competitive inhibitor of HCMV protein kinase pUL97 that blocks viral replication.
potent and selective ATP-competitive inhibitor of HCMV protein kinase pUL97; orally available antiviral agent

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SML2455SML2825SML2826
description

≥98% (HPLC)

description

≥98% (HPLC)

description

≥98% (HPLC)

description

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

form

powder

form

powder

form

powder

form

powder

Quality Level

100

Quality Level

-

Quality Level

100

Quality Level

100

storage temp.

−20°C

storage temp.

−20°C

storage temp.

2-8°C

storage temp.

2-8°C

solubility

DMSO: 2 mg/mL, clear

solubility

DMSO: 2 mg/mL, clear

solubility

DMSO: 2 mg/mL, clear

solubility

DMSO: 2 mg/mL, clear

color

white to beige

color

white to beige

color

white to beige

color

white to beige


WGK

WGK 3

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Codes

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library




Questions

Reviews

No rating value

Active Filters