Skip to Content
Merck
All Photos(1)

Key Documents

SML2940

Sigma-Aldrich

ML417

≥98% (HPLC)

Synonym(s):

(1H-Indol-2-yl)(4-(2-(4-methoxyphenoxy)ethyl)piperazin-1-yl)methanone, Methanone, 1H-indol-2-yl[4-[2-(4-methoxyphenoxy)ethyl]-1-piperazinyl]-

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€65.90
25 MG
€267.00

€65.90


Estimated to ship on25 April 2025


Request a Bulk Order

Select a Size

Change View
5 MG
€65.90
25 MG
€267.00

About This Item

Empirical Formula (Hill Notation):
C22H25N3O3
CAS Number:
Molecular Weight:
379.45
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

€65.90


Estimated to ship on25 April 2025


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

N2(CCN(CC2)C(=O)c3[nH]c4c(c3)cccc4)CCOc1ccc(cc1)OC

InChI key

HAZPAMUWUHDPDA-UHFFFAOYSA-N

Biochem/physiol Actions

ML417 is a blood-brain barrier permeable, potent and highly selective agonist of D3 dopamine receptor (D3R) with very limited cross-reactivity at other GPCRs. ML417 potently induces G-protein activation, D3R-mediated b-arrestin translocation and pERK phosphorylation. It does not activate other dopamine receptors. ML417 protect D3R-expressing dopaminergic neurons from 6-OHDA induced cell death.
blood-brain barrier permeable, potent and highly selective agonist of D3 dopamine receptor (D3R)

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Amy E Moritz et al.
Journal of medicinal chemistry, 63(10), 5526-5567 (2020-04-29)
To identify novel D3 dopamine receptor (D3R) agonists, we conducted a high-throughput screen using a β-arrestin recruitment assay. Counterscreening of the hit compounds provided an assessment of their selectivity, efficacy, and potency. The most promising scaffold was optimized through medicinal

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service