Skip to Content
Merck
All Photos(1)

Key Documents

SML2927

Sigma-Aldrich

H3B-193

≥98% (HPLC)

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€74.80
25 MG
€302.00

€74.80


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
5 MG
€74.80
25 MG
€302.00

About This Item

Empirical Formula (Hill Notation):
C19H23FN4O2S
CAS Number:
Molecular Weight:
390.47
UNSPSC Code:
12352200
NACRES:
NA.77

€74.80


Estimated to ship on28 May 2025


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C(NC1=NC(C)=CS1)C2CCN(C(N(CC3=CC=C(F)C=C3)C)=O)CC2

InChI

1S/C19H23FN4O2S/c1-13-12-27-18(21-13)22-17(25)15-7-9-24(10-8-15)19(26)23(2)11-14-3-5-16(20)6-4-14/h3-6,12,15H,7-11H2,1-2H3,(H,21,22,25)

InChI key

FYQIXROHNRZVMM-UHFFFAOYSA-N

Biochem/physiol Actions

H3B-193 is a cell penetrant, potent and selective allosteric inhibitor of the carbamoyl phosphate synthetase 1 (CPS1) that blocks ATP hydrolysis in the first step of carbamoyl phosphate synthesis
cell penetrant, potent and selective allosteric inhibitor of the carbamoyl phosphate synthetase 1 (CPS1)

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shihua Yao et al.
Cell chemical biology, 27(3), 259-268 (2020-02-06)
Carbamoyl phosphate synthetase 1 (CPS1) catalyzes the first step in the ammonia-detoxifying urea cycle, converting ammonia to carbamoyl phosphate under physiologic conditions. In cancer, CPS1 overexpression supports pyrimidine synthesis to promote tumor growth in some cancer types, while in others

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service