Skip to Content
Merck
All Photos(1)

Key Documents

SML2892

Sigma-Aldrich

LY2584702 tosylate

≥98% (HPLC)

Synonym(s):

4-[4-[4-[4-Fluoro-3-(trifluoromethyl)phenyl]-1-methyl-1H-imidazol-2-yl]-1-piperidinyl]-1H-pyrazolo[3,4-d]pyrimidine, 4-methylbenzenesulfonate (1:1), LY 2584702 tosylate, LY 2779964, LY-2584702 tosylate, LY-2779964, LY2779964, LYS6K2 tosylate

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€73.00
25 MG
€237.00

€73.00


Estimated to ship on29 May 2025


Request a Bulk Order

Select a Size

Change View
5 MG
€73.00
25 MG
€237.00

About This Item

Empirical Formula (Hill Notation):
C21H19F4N7 · C7H8O3S
CAS Number:
Molecular Weight:
617.62
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

€73.00


Estimated to ship on29 May 2025


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

FC(F)(F)c2c(ccc(c2)c3nc([n](c3)C)C4CCN(CC4)c5ncnc6n[nH]cc65)F.[S](=O)(=O)(O)c1ccc(cc1)C

InChI

1S/C21H19F4N7.C7H8O3S/c1-31-10-17(13-2-3-16(22)15(8-13)21(23,24)25)29-19(31)12-4-6-32(7-5-12)20-14-9-28-30-18(14)26-11-27-20;1-6-2-4-7(5-3-6)11(8,9)10/h2-3,8-12H,4-7H2,1H3,(H,26,27,28,30);2-5H,1H3,(H,8,9,10)

InChI key

HDYUXDNMHBQKAU-UHFFFAOYSA-N

Biochem/physiol Actions

LY2584702 is an orally active, ATP-competitive, potent and selective ribosomal protein S6 kinase (p70 S6 kinase, p70S6K, S6K1) inhibitor (IC50 = 4 nM; selective over 83 other kinases and 45 cell surface markers). LY2584702 inhibits HCT116 cellular S6 phosphorylation (IC50 0.1-0.24 μM) and shows anti-tumor efficacy in U87MG glioblastoma and HCT116 colon carcinoma xenograft models in vivo (2.5 mg/kg bid. po.).
Orally active, ATP-competitive, potent and selective ribosomal protein S6 kinase (p70 S6 kinase, p70S6K, S6K1) inhibitor with in vivo anti-tumor efficacy.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anthony Tolcher et al.
European journal of cancer (Oxford, England : 1990), 50(5), 867-875 (2014-01-21)
LY2584702 tosylate (hereafter referred to as LY2584702) is a potent, highly selective adenosine triphosphate (ATP) competitive inhibitor against p70 S6 kinase, a downstream component of the phosphatidylinositol-3-kinase signalling pathway which regulates cell proliferation and survival. LY2584702 exhibited anti-tumour activity in
Honyin Chiu et al.
Journal of immunology (Baltimore, Md. : 1950), 202(2), 579-590 (2018-12-12)
During an adaptive immune response, activated mature B cells give rise to Ab-secreting plasma cells to fight infection. B cells undergo Ab class switching to produce different classes of Abs with varying effector functions. The mammalian/mechanistic target of rapamycin (mTOR)
Anne-Sophie Archambault et al.
PloS one, 12(1), e0169804-e0169804 (2017-01-10)
LTB4 is an inflammatory lipid mediator mainly biosynthesized by leukocytes. Since its implication in inflammatory diseases is well recognized, many tools to regulate its biosynthesis have been developed and showed promising results in vitro and in vivo, but mixed results
Valentina Romeo et al.
Journal of immunology (Baltimore, Md. : 1950), 203(1), 137-147 (2019-05-17)
PI3K is one of the most frequently mutated genes in cancers and has been the target of numerous anticancer therapies. With the additional development of therapeutics that mobilize the immune system, such as Abs with effector functions, bispecific Abs, and
Elizabeth K K Glennon et al.
Cell reports, 26(12), 3391-3399 (2019-03-21)
Plasmodium parasites are highly selective when infecting hepatocytes and induce many changes within the host cell upon infection. While several host cell factors have been identified that are important for liver infection, our understanding of what facilitates the maintenance of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service