Skip to Content
Merck
All Photos(1)

Key Documents

SML2887

Sigma-Aldrich

Firocoxib

≥98% (HPLC)

Synonym(s):

3-(Cyclopropyl-methoxy)-5,5-Dimethy l-4-(4-methylsulfonyl)phenyl)-5H-furan-2-one, 3-(Cyclopropylmethoxy)-5,5-dimethyl-4-(4-(methylsulfonyl)phenyl)furan-2(5H)-one, 3-(Cyclopropylmethoxy)-5,5-dimethyl-4-[4-(methylsulfonyl)phenyl]-2(5H)-furanone, ML 1785713, Previcox, 3-Cyclopropylmethoxy-4-(4-methanesulfonylphenyl)-5,5-dimethyl-5H-furan-2-one

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€85.90
25 MG
€349.00

€85.90


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
€85.90
25 MG
€349.00

About This Item

Empirical Formula (Hill Notation):
C17H20O5S
CAS Number:
Molecular Weight:
336.40
Beilstein:
8335837
MDL number:
UNSPSC Code:
51111800
NACRES:
NA.77

€85.90


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=S(C1=CC=C(C2=C(OCC3CC3)C(OC2(C)C)=O)C=C1)(C)=O

InChI

1S/C17H20O5S/c1-17(2)14(12-6-8-13(9-7-12)23(3,19)20)15(16(18)22-17)21-10-11-4-5-11/h6-9,11H,4-5,10H2,1-3H3

InChI key

FULAPETWGIGNMT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Firocoxib is an orally available non-steroidal anti-inflammatory drug (NSAID) that selectively and potently inhibits cyclooxygenase-2-(COX-2). Firocoxib is approved to use in dogs and horses.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Amy K Stuart et al.
Journal of veterinary pharmacology and therapeutics, 42(6), 640-646 (2019-08-23)
Pharmacokinetic (PK) studies of oral firocoxib in large animal species have been limited to horses, preruminating calves, and adult camels. The aim of this study was to describe pharmacokinetics and bioavailability of firocoxib in adult goats. Ten healthy adult goats
Milena M Madry et al.
BMC veterinary research, 12, 84-84 (2016-06-03)
Compared to blood or urine, drugs can be detected for much longer periods in the long hair of horses. The aim of this study was to establish and validate a highly sensitive liquid chromatography tandem mass spectrometry (LC-MS/MS) method for
Prangtip Phuwapattanachart et al.
Journal of veterinary science, 18(2), 175-182 (2016-08-16)
The postoperative analgesic effects of firocoxib in ovariohysterectomized cats were observed. Twenty-four cats were divided into 3 groups: control (no medicine), firocoxib-1 (1 mg/kg/day) and firocoxib-3 (3 mg/kg/day). Colorado pain scale scores (CPSS), composite pain scores (CPS), and buccal mucosal

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service