Skip to Content
Merck
All Photos(1)

Key Documents

SML2854

Sigma-Aldrich

Polmacoxib

≥98% (HPLC)

Synonym(s):

4-[3-(3-fluorophenyl)-4,5-dihydro-5,5-dimethyl-4-oxo-2-furanyl]-benzenesulfonamide, CG100649

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€73.00
25 MG
€237.00

€73.00


Check Cart for Availability


Select a Size

Change View
5 MG
€73.00
25 MG
€237.00

About This Item

Empirical Formula (Hill Notation):
C18H16FNO4S
CAS Number:
Molecular Weight:
361.39
MDL number:
UNSPSC Code:
51111800
NACRES:
NA.77

€73.00


Check Cart for Availability

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

NS(C1=CC=C(C=C1)C(OC2(C)C)=C(C2=O)C3=CC=CC(F)=C3)(=O)=O

InChI

1S/C18H16FNO4S/c1-18(2)17(21)15(12-4-3-5-13(19)10-12)16(24-18)11-6-8-14(9-7-11)25(20,22)23/h3-10H,1-2H3,(H2,20,22,23)

InChI key

IJWPAFMIFNSIGD-UHFFFAOYSA-N

Biochem/physiol Actions

Inhibitor of cyclooxygenase 2 (COX-2) and the carbonic anhydrases CAI and CAII
Polmacoxib is a nonsteroidal anti-inflammatory drug (NSAID) that acts as an inhibitor of cyclooxygenase 2 (COX-2) and the carbonic anhydrases CAI and CAII. It has also been found to inhibit colorectal adenoma and tumor growth in mouse models.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hyun Tae Kim et al.
Biochemical and biophysical research communications, 478(1), 1-6 (2016-08-01)
Polmacoxib is not only a selective COX-2 inhibitor but also a potent inhibitor of carbonic anhydrases (CAs). Both CA I and CA II are highly expressed in the GI tract and kidneys, organs that are also thought to be the
Myungchul Lee et al.
Clinics in orthopedic surgery, 9(4), 439-457 (2017-12-05)
The aim of this study was to evaluate the safety and analgesic efficacy of polmacoxib 2 mg versus placebo in a superiority comparison or versus celecoxib 200 mg in a noninferiority comparison in patients with osteoarthritis (OA). This study was

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service