Skip to Content
Merck
All Photos(1)

Key Documents

SML2791

Sigma-Aldrich

SBI-797812

≥98% (HPLC)

Synonym(s):

1-[4-(8-Oxa-3-azabicyclo[3.2.1]octane-3-sulfonyl)-phenyl]-3-pyridin-4-ylmethylurea, N-[4-(8-Oxa-3-azabicyclo[3.2.1]oct-3-ylsulfonyl)phenyl]-N′-(4-pyridinylmethyl)-urea

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€85.90
25 MG
€368.00

€85.90


Estimated to ship on26 May 2025


Request a Bulk Order

Select a Size

Change View
5 MG
€85.90
25 MG
€368.00

About This Item

Empirical Formula (Hill Notation):
C19H22N4O4S
CAS Number:
Molecular Weight:
402.47
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

€85.90


Estimated to ship on26 May 2025


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C(NC1=CC=C(S(=O)(N(C2)C[C@H]3O[C@@H]2CC3)=O)C=C1)NCC4=CC=NC=C4

InChI

1S/C19H22N4O4S/c24-19(21-11-14-7-9-20-10-8-14)22-15-1-5-18(6-2-15)28(25,26)23-12-16-3-4-17(13-23)27-16/h1-2,5-10,16-17H,3-4,11-13H2,(H2,21,22,24)/t16-,17+

InChI key

KTSOHNHLOLGQCY-CALCHBBNSA-N

Biochem/physiol Actions

SBI-797812 is a cell penetrant and potent activator of nicotinamide phosphoribosyltransferase (NAMPT) that increases intracellular levels of NMN and NAD+ in cells. SBI-797812 blocks binding of inhibitors to NAMPT and shifts the reaction equilibrium towards NMN formation. It elevates levels of NAD+ in mice liver.
cell penetrant and potent activator of NAMPT increases intracellular levels of NMN and NAD+ in cells

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stephen J Gardell et al.
Nature communications, 10(1), 3241-3241 (2019-07-22)
Pharmacological strategies that boost intracellular NAD+ are highly coveted for their therapeutic potential. One approach is activation of nicotinamide phosphoribosyltransferase (NAMPT) to increase production of nicotinamide mononucleotide (NMN), the predominant NAD+ precursor in mammalian cells. A high-throughput screen for NAMPT activators

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service