Skip to Content
Merck
All Photos(1)

Key Documents

SML2703

Sigma-Aldrich

BAY-293

≥98% (HPLC)

Synonym(s):

6,7-Dimethoxy-2-methyl-N-[(R)-1-(4-{2-[(methylamino)methyl]phenyl}thien-2-yl)ethyl]quinazolin-4-amine, BAY 293, BAY293

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€106.00
25 MG
€428.00

€106.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
€106.00
25 MG
€428.00

About This Item

Empirical Formula (Hill Notation):
C25H28N4O2S
CAS Number:
Molecular Weight:
448.58
UNSPSC Code:
12352200
NACRES:
NA.77

€106.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

optical activity

[α]/D -85.0 to -103.0°, c = 1.0 in DMSO

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

C[C@H](C1=CC(C2=CC=CC=C2CNC)=CS1)NC3=C4C=C(C(OC)=CC4=NC(C)=N3)OC

InChI

1S/C25H28N4O2S/c1-15(24-10-18(14-32-24)19-9-7-6-8-17(19)13-26-3)27-25-20-11-22(30-4)23(31-5)12-21(20)28-16(2)29-25/h6-12,14-15,26H,13H2,1-5H3,(H,27,28,29)/t15-/m1/s1

InChI key

WEGLOYDTDILXDA-OAHLLOKOSA-N

Biochem/physiol Actions

BAY-293 selectively inhibits SOS1-mediated KRAS activation (IC50 = 52 nM; 50 nM wt or G12C hKRAS & 10 nM SOS1cat) via direct SOS1 binding, blocking SOS1-KRAS interaction (IC50 = 21 nM; 10 nM hSOS1cat & 5 nM hKRAS G12C) without affecting (IC50 >20 μM) CRAF RBD-KRAS interaction, KRAS G12C activation by SOS2, Cdc42 activation by DBS, or EGFR kinase activity. BAY-293 downregulates cellular active RAS (IC50 = 200 nM/Calu-1, 410 nM/HeLa) and downstream ERK phosphorylation (K-562, 150 nM/Calu-1) with good target selectivity as assessed over 358 kinases (<37% inhibition at 1 μM) and a panel of 77 GPCRs, transporters, nuclear receptors and enzymes.
Selective inhibitor against SOS1-mediated KRAS activation without affecting CRAF RBD-KRAS interaction or KRAS G12C activation by SOS2.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Roman C Hillig et al.
Proceedings of the National Academy of Sciences of the United States of America, 116(7), 2551-2560 (2019-01-27)
Since the late 1980s, mutations in the RAS genes have been recognized as major oncogenes with a high occurrence rate in human cancers. Such mutations reduce the ability of the small GTPase RAS to hydrolyze GTP, keeping this molecular switch

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service