Skip to Content
Merck
All Photos(1)

Key Documents

SML2699

Sigma-Aldrich

SHMT inhibitor Hit 1

≥98% (HPLC)

Synonym(s):

4,4′-[2,4-Pyrimidinediylbis(4,1-phenyleneimino)]bis[4-oxo-butanoic acid

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€89.30
25 MG
€361.00

€89.30


Estimated to ship on01 June 2025


Request a Bulk Order

Select a Size

Change View
5 MG
€89.30
25 MG
€361.00

About This Item

Empirical Formula (Hill Notation):
C24H22N4O6
CAS Number:
Molecular Weight:
462.45
UNSPSC Code:
12352200
NACRES:
NA.77

€89.30


Estimated to ship on01 June 2025


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C(NC1=CC=C(C2=NC=CC(C3=CC=C(C=C3)NC(CCC(O)=O)=O)=N2)C=C1)CCC(O)=O

InChI

1S/C24H22N4O6/c29-20(9-11-22(31)32)26-17-5-1-15(2-6-17)19-13-14-25-24(28-19)16-3-7-18(8-4-16)27-21(30)10-12-23(33)34/h1-8,13-14H,9-12H2,(H,26,29)(H,27,30)(H,31,32)(H,33,34)

InChI key

LZRQXDAEFLBXCL-UHFFFAOYSA-N

Biochem/physiol Actions

Inhibition of serine/glycine synthesis by SHMT inhibitors can be an effective strategy to treat serine/glycine synthesis-dependent cancers.[1] Therefore, SHMT inhibitor can be a potential cancer drug target.[2] SHMT inhibitor Hit 1 may inhibit SHMT with an IC50 of 0.53 μM.[2]
SHMT inhibitor Hit 1 is a potent and specific inhibitor of serine hydroxymethyltransferase (SHMT). SHMT inhibitor Hit 1 potently inhibits SHMT1 and SHMT2 in rat liver homogenate.
potent and specific inhibitor of serine hydroxymethyltransferase (SHMT)

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shauni Lien Geeraerts et al.
Molecular cancer therapeutics, 20(1), 50-63 (2020-11-19)
Metabolic rewiring is a hallmark of cancer that supports tumor growth, survival, and chemotherapy resistance. Although normal cells often rely on extracellular serine and glycine supply, a significant subset of cancers becomes addicted to intracellular serine/glycine synthesis, offering an attractive

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service