Skip to Content
Merck
All Photos(1)

Key Documents

SML2687

Sigma-Aldrich

dBET1

≥98% (HPLC)

Synonym(s):

(6S)-4-(4-Chlorophenyl)-N-[4-[[2-[[2-(2,6-dioxo-3-piperidinyl)-2,3-dihydro-1,3-dioxo-1H-isoindol-4-yl]oxy]acetyl]amino]butyl]-2,3,9-trimethyl-6H-thieno[3,2-f][1,2,4]triazolo[4,3-a][1,4]diazepine-6-acetamide, 2-((S)-4-(4-Chlorophenyl)-2,3,9-trimethyl-6H-thieno[3,2-f][1,2,4]triazolo[4,3-a][1,4]diazepin-6-yl)-N-(4-(2-(2-(2,6-dioxopiperidin-3-yl)-1,3-dioxoisoindolin-4-yloxy)acetamido)butyl)acetamide

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€44.20
5 MG
€180.00

€44.20


Estimated to ship on14 April 2025


Request a Bulk Order

Select a Size

Change View
1 MG
€44.20
5 MG
€180.00

About This Item

Empirical Formula (Hill Notation):
C38H37ClN8O7S
CAS Number:
Molecular Weight:
785.27
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

€44.20


Estimated to ship on14 April 2025


Request a Bulk Order

ligand

thalidomide

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

[s]1c2c(c(c1C)C)C(=N[C@H](c7[n]2c(nn7)C)CC(=O)NCCCCNC(=O)COc4c5c(ccc4)C(=O)N(C5=O)C6CCC(=O)NC6=O)c3ccc(cc3)Cl

InChI key

LKEGXJXRNBALBV-PMCHYTPCSA-N

Biochem/physiol Actions

A JQ1-and-phthalimide conjugate that causes the degradation of BRD4 protein
dBET1 is a JQ1-and-phthalimide conjugate that causes the degradation of BRD4 protein, a transcriptional coactivator that regulates the expression of genes that promote cancer cell proliferation and survival. JQ1 is a selective BET bromodomain (BRD) inihbitor. The phthalimide portion is a derivative of the drug thalidomide, which is known to bind to cereblon (CRBN), part of an ubiquitin E3 ligase complex resulting in ubiquitylation and degradation by the proteasome. The conjugate dBET1 induced highly selective cereblon-dependent BET protein degradation both in vitro and in vivo. It induced complete CRBN-dependent proteosomal degradation of BRD4 in a human acute myeloid leukaemia (AML) cell line and delayed leukemia progression in mice.

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Afua A Akuffo et al.
The Journal of biological chemistry, 293(16), 6187-6200 (2018-02-17)
Upon binding to thalidomide and other immunomodulatory drugs, the E3 ligase substrate receptor cereblon (CRBN) promotes proteosomal destruction by engaging the DDB1-CUL4A-Roc1-RBX1 E3 ubiquitin ligase in human cells but not in mouse cells, suggesting that sequence variations in CRBN may
Georg E Winter et al.
Science (New York, N.Y.), 348(6241), 1376-1381 (2015-05-23)
The development of effective pharmacological inhibitors of multidomain scaffold proteins, notably transcription factors, is a particularly challenging problem. In part, this is because many small-molecule antagonists disrupt the activity of only one domain in the target protein. We devised a
Kelly M DeMars et al.
Neurochemistry international, 127, 94-102 (2019-03-16)
Neuroinflammation after stroke significantly contributes to neuronal cell death. Bromodomain and Extra Terminal Domain (BET) proteins are essential to inflammatory gene transcription. BET proteins (BRD2, BRD3, BRD4, and BRDT) have varied effects including chromatin remodeling, histone acetyltransferase activity, and as

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service