Skip to Content
Merck
All Photos(1)

Key Documents

SML2681

Sigma-Aldrich

cRIPGBM

≥98% (HPLC)

Synonym(s):

1-Benzyl-3-(4-fluorobenzyl)-2-methyl-4,9-dioxo-4,9-dihydro-1H-naphtho[2,3-d]imidazol-3-ium chloride, RIPGBM proapoptic metabolite

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€68.20
25 MG
€286.00

€68.20


Estimated to ship on25 May 2025


Request a Bulk Order

Select a Size

Change View
5 MG
€68.20
25 MG
€286.00

About This Item

Empirical Formula (Hill Notation):
C26H20ClFN2O2
CAS Number:
Molecular Weight:
446.90
UNSPSC Code:
12352200
NACRES:
NA.77

€68.20


Estimated to ship on25 May 2025


Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C1C2=C([N+](CC3=CC=C(F)C=C3)=C(C)N2CC4=CC=CC=C4)C(C5=C1C=CC=C5)=O.[Cl-]

InChI

1S/C26H20FN2O2.ClH/c1-17-28(15-18-7-3-2-4-8-18)23-24(29(17)16-19-11-13-20(27)14-12-19)26(31)22-10-6-5-9-21(22)25(23)30;/h2-14H,15-16H2,1H3;1H/q+1;/p-1

InChI key

IHNGETPFBJHWFE-UHFFFAOYSA-M

Biochem/physiol Actions

Apoptosis inducer that binds receptor-interacting protein kinase 2 (RIPK2, RIP2) and promotes the proapoptotic RIPK2/caspase-1 complex formation.
cRIPGBM is an apoptosis inducer (GBM-1/-5/-39 EC50 = 63/95/290 nM; human neural progenitor cells (NPCs)/astrocytes/lung fibroblasts EC50 = 190/710/960 nM) that binds receptor-interacting protein kinase 2 (RIPK2, RIP2) and promotes the formation of a proapoptotic RIPK2/caspase-1 complex. Cell type-selective metabolic redox conversion of RIPGBM to cRIPGBM in glioblastoma multiforme (GBM) cancer stem cells (CSCs) form the basis of selective GBM CSCs apoptosis induction by RIPGBM over non-GBM cultures.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Natasha C Lucki et al.
Proceedings of the National Academy of Sciences of the United States of America, 116(13), 6435-6440 (2019-03-09)
Glioblastoma multiforme (GBM; grade IV astrocytoma) is the most prevalent and aggressive form of primary brain cancer. A subpopulation of multipotent cells termed GBM cancer stem cells (CSCs) play a critical role in tumor initiation, tumor maintenance, metastasis, drug resistance

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service