Skip to Content
Merck
All Photos(1)

Key Documents

SML2673

Sigma-Aldrich

Deferasirox

≥98% (HPLC)

Synonym(s):

4-[3,5-bis(2-Hydroxyphenyl)-1H-1,2,4-triazol-1-yl]benzoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H15N3O4
CAS Number:
Molecular Weight:
373.36
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

N2(N\C(=C4\C=CC=CC\4=O)\N\C\2=C3\C=CC=CC\3=O)c1ccc(cc1)C(=O)O

InChI

1S/C21H15N3O4/c25-17-7-3-1-5-15(17)19-22-20(16-6-2-4-8-18(16)26)24(23-19)14-11-9-13(10-12-14)21(27)28/h1-12,22-23H,(H,27,28)/b19-15-,20-16+

InChI key

FMSOAWSKCWYLBB-VBGLAJCLSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Deferasirox has been used as an iron chelator to test its effect on clofazimine mediated growth inhibition and rescue in Salmonella typhimurium.

Biochem/physiol Actions

Deferasirox belongs to the N-substituted bis-hydroxyphenyl-triazole family of tridentate iron chelators.
Deferasirox is an orally available iron chelator used clinically for reduction of chronic iron overload in diseases such as β-thalassemia.
Orally available iron chelator

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Goldie Y L Lui et al.
Oncotarget, 6(22), 18748-18779 (2015-07-01)
Newer and more potent therapies are urgently needed to effectively treat advanced cancers that have developed resistance and metastasized. One such strategy is to target cancer cell iron metabolism, which is altered compared to normal cells and may facilitate their
Kangna Cao et al.
European journal of clinical pharmacology, 76(1), 51-59 (2019-11-05)
Our aim was to evaluate the influence of genetic polymorphisms involved in the metabolism and transportation of deferasirox on deferasirox pharmacokinetics in the Chinese population. Thirty-eight healthy Chinese subjects were administered with a single dose of 20 mg kg-1 deferasirox.
Juan Daniel Díaz-García et al.
Nature reviews. Nephrology, 10(10), 574-586 (2014-07-23)
In 2005, the oral iron chelator deferasirox was approved by the FDA for clinical use as a first-line therapy for blood-transfusion-related iron overload. Nephrotoxicity is the most serious and frequent adverse effect of deferasirox treatment. This nephrotoxicity can present as
Omid Reza Zekavat et al.
Journal of pediatric hematology/oncology, 43(1), e26-e28 (2020-09-15)
This study was performed on patients with transfusion-dependent beta-thalassemia (TDT) to investigate the effect of HFE gene mutations of iron overload in a large group of patients with TDT major and its relationship with heart and liver T2* magnetic resonance
Abbas Rahdar et al.
Life sciences, 270, 119146-119146 (2021-02-06)
Deferasirox (DFX) was formulated into oil-in-water microemulsions in the presence of pluronicto improve its oral bioavailability. The size of the DFX-loadedmicroemulsions system measured by dynamic light scattering (DLS) was about 9 nm. The anti-proliferative and anti-lipid peroxidation effects of DFX and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service