Skip to Content
Merck
All Photos(1)

Key Documents

SML2669

Sigma-Aldrich

ITK7

≥98% (HPLC)

Synonym(s):

8-methyl-7-(prop-1-yn-1-yl)-2-((pyrimidin-2-ylthio)methyl)quinazolin-4(3H)-one

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€112.00
25 MG
€359.00

€112.00


Estimated to ship on17 May 2025


Request a Bulk Order

Select a Size

Change View
5 MG
€112.00
25 MG
€359.00

About This Item

Empirical Formula (Hill Notation):
C17H14N4OS
CAS Number:
Molecular Weight:
322.38
UNSPSC Code:
12352200
NACRES:
NA.77

€112.00


Estimated to ship on17 May 2025


Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

CC1=C(N=C(CSC2=NC=CC=N2)NC3=O)C3=CC=C1C#CC

Biochem/physiol Actions

ITK7 is a cell penetrant, potent and selective inhibitor of PARP11 mono-ADP-ribosylation (MARylation) activity.
cell penetrant, potent and selective inhibitor of PARP11 mono-ADP-ribosylation

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ilsa T Kirby et al.
Cell chemical biology, 25(12), 1547-1553 (2018-10-23)
Poly-ADP-ribose polymerases (PARPs1-16) play pivotal roles in diverse cellular processes. PARPs that catalyze poly-ADP-ribosylation (PARylation) are the best characterized PARP family members because of the availability of potent and selective inhibitors for these PARPs. There has been comparatively little success in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service