Skip to Content
Merck
All Photos(1)

Key Documents

SML2637

Sigma-Aldrich

NPD8733

≥98% (HPLC)

Synonym(s):

N-[2-(4-Methoxybenzoyl)-3-benzofuranyl]acetamide

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€71.60
25 MG
€299.00

€71.60


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
€71.60
25 MG
€299.00

About This Item

Empirical Formula (Hill Notation):
C18H15NO4
CAS Number:
Molecular Weight:
309.32
UNSPSC Code:
12352200
NACRES:
NA.77

€71.60


Check Cart for Availability

Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

N(c1c2c([o]c1C(=O)c3ccc(cc3)OC)cccc2)C(=O)C

InChI

1S/C18H15NO4/c1-11(20)19-16-14-5-3-4-6-15(14)23-18(16)17(21)12-7-9-13(22-2)10-8-12/h3-10H,1-2H3,(H,19,20)

InChI key

NQZBUYPGHZTWIR-UHFFFAOYSA-N

Biochem/physiol Actions

NPD8733 is an inhibitor of cancer cell-enhanced fibroblast migration. NPD8733 binds to valosin-containing protein (VCP)/p97.
inhibitor of cancer cell-enhanced fibroblast migration

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kruthi Suvarna et al.
The Journal of biological chemistry, 294(9), 2988-2996 (2019-01-06)
Carcinoma-associated fibroblasts are fibroblasts activated by surrounding cancer cells. Carcinoma-associated fibroblasts exhibit enhanced cell migration, which plays an important role in cancer metastasis. Previously, we demonstrated enhanced migration of NIH3T3 fibroblasts when they were cultured in the presence of MCF7

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service