Skip to Content
Merck
All Photos(1)

Key Documents

SML2630

Sigma-Aldrich

Intronistat B hydrobromide

≥98% (HPLC)

Synonym(s):

N-(2-(Pyrrolidin-1-yl)ethyl)-2-(3,4,5-trihydroxybenzoyl) benzofuran-5-carboxamide hydrobromide

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€73.00
25 MG
€297.00

€73.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
€73.00
25 MG
€297.00

About This Item

Empirical Formula (Hill Notation):
C22H22N2O6 · xHBr
Molecular Weight:
410.42 (free base basis)
UNSPSC Code:
51111800
NACRES:
NA.77

€73.00


Check Cart for Availability

Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to dark gray

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

Biochem/physiol Actions

Inhibitor of the self-splicing group IIB intron found in fungi
Intronistat B is a potent inhibitor of the self-splicing group IIB intron, a mitochondrial RNA tertiary structure necessary for respiration in fungi, yeast and plants, but not found in vertebrates. Intronistat B was found to have antifungal activity comparable to that of amphotericin B in inhibiting growth of the Candida parapsilosis. It did not inhibit spliceosomal processing, group I intron splicing or splicing of group II introns from subclasses other than subclass IIB and was not toxic to human cells.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Olga Fedorova et al.
Nature chemical biology, 14(12), 1073-1078 (2018-10-17)
Specific RNA structures control numerous metabolic processes that impact human health, and yet efforts to target RNA structures de novo have been limited. In eukaryotes, the self-splicing group II intron is a mitochondrial RNA tertiary structure that is absent in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service