Skip to Content
Merck
All Photos(1)

Key Documents

SML2626

Sigma-Aldrich

PEP87 trifluoroacetate salt

≥95% (HPLC)

Synonym(s):

Trp-Ser-Tyr-Asn-Ala-Glu-Leu-Leu-Val-Ala-Met-Glu-Asn-Gln-His-Thr-Ile trifluoroacetate salt, WSYNAELLVAMENQHTI trifluoroacetate salt

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€128.00

€128.00


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
1 MG
€128.00

About This Item

Empirical Formula (Hill Notation):
C90H135N23O28S · xC2HF3O2
Molecular Weight:
2019.24 (free base basis)
UNSPSC Code:
12352200
NACRES:
NA.77

€128.00


Estimated to ship on28 May 2025


Request a Bulk Order

Assay

≥95% (HPLC)

form

film

storage condition

desiccated

color

white to beige

storage temp.

−20°C

Biochem/physiol Actions

PEP87, a highly conserved helical fragment of hemagglutinin HA2 subunit, is an inhibitor of H7 and H5 hemagglutinin (HA) mediated influenza virus entry. PEP87 binds to the hemagglutinin trimer where it disrupts native tertiary structure and prevents HA function.
inhibitor of H7 and H5 hemagglutinin mediated influenza virus entry

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service