Skip to Content
Merck
All Photos(1)

Key Documents

SML2609

Sigma-Aldrich

JH-I-25

≥98% (HPLC)

Synonym(s):

N-[2-Methoxy-4-(4-morpholinyl)phenyl]-6-(1H-pyrazol-3-yl)-2-pyridinecarboxamide

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€117.00
25 MG
€375.00

€117.00


Estimated to ship on29 March 2025


Request a Bulk Order

Select a Size

Change View
5 MG
€117.00
25 MG
€375.00

About This Item

Empirical Formula (Hill Notation):
C20H21N5O3
CAS Number:
Molecular Weight:
379.41
UNSPSC Code:
12352200
NACRES:
NA.77

€117.00


Estimated to ship on29 March 2025


Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear (warmed)

storage temp.

2-8°C

SMILES string

O=C(NC1=C(C=C(C=C1)N2CCOCC2)OC)C3=NC(C4=NNC=C4)=CC=C3

InChI

1S/C20H21N5O3/c1-27-19-13-14(25-9-11-28-12-10-25)5-6-17(19)23-20(26)18-4-2-3-15(22-18)16-7-8-21-24-16/h2-8,13H,9-12H2,1H3,(H,21,24)(H,23,26)

Biochem/physiol Actions

JH-I-25 is a potent and selective type I inhibitor of IRAK1 and IRAK4 (interleukin-1 receptor–associated kinase 1 and 4). JH-I-25 binds preferentially to the unphosphorylated the ATP-site of the kinase in its active, DFG-in, state.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Li Wang et al.
The Journal of biological chemistry, 294(12), 4511-4519 (2019-01-27)
Interleukin-1 receptor-associated kinase 4 (IRAK4) is a key player in innate immune and inflammatory responses, performing a critical role in signal transduction downstream of Toll-like receptors and interleukin-1 (IL-1) receptors. Upon ligand binding and via its N-terminal death domain, IRAK4
Li Wang et al.
Proceedings of the National Academy of Sciences of the United States of America, 114(51), 13507-13512 (2017-12-07)
Interleukin 1 (IL-1) receptor-associated kinases (IRAKs) are serine/threonine kinases that play critical roles in initiating innate immune responses against foreign pathogens and other types of dangers through their role in Toll-like receptor (TLR) and interleukin 1 receptor (IL-1R) mediated signaling
George M Buckley et al.
Bioorganic & medicinal chemistry letters, 18(11), 3211-3214 (2008-05-14)
The synthesis and profile of a series of amides are described. Some of these compounds were potent IRAK-4 inhibitors and two examples were evaluated in vivo.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service