Skip to Content
Merck
All Photos(1)

Key Documents

SML2595

Sigma-Aldrich

AZD1208

≥98% (HPLC)

Synonym(s):

(5Z)-5-[[2-[(3R)-3-Amino-1-piperidinyl][1,1′-biphenyl]-3-yl]methylene]-2,4-thiazolidinedione, (R,Z)-5-((2-(3-Aminopiperidin-1-yl)biphenyl-3-yl)methylene)thiazolidine-2,4-dione, AZD 1208, AZD-1208

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€63.10
25 MG
€255.00

€63.10


Estimated to ship on29 May 2025


Request a Bulk Order

Select a Size

Change View
5 MG
€63.10
25 MG
€255.00

About This Item

Empirical Formula (Hill Notation):
C21H21N3O2S
CAS Number:
Molecular Weight:
379.48
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

€63.10


Estimated to ship on29 May 2025


Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

S1\C(=C/c2c(c(ccc2)c4ccccc4)N3C[C@@H](CCC3)N)\C(=O)NC1=O

InChI

1S/C21H21N3O2S/c22-16-9-5-11-24(13-16)19-15(12-18-20(25)23-21(26)27-18)8-4-10-17(19)14-6-2-1-3-7-14/h1-4,6-8,10,12,16H,5,9,11,13,22H2,(H,23,25,26)/b18-12-/t16-/m1/s1

InChI key

MCUJKPPARUPFJM-UWCCDQBKSA-N

Biochem/physiol Actions

AZD1208 is an orally available, potent ATP-competitive pan-Pim kinase inhibitor (Pim-1/-2/-3 Ki = 0.1/1.92/0.4 nM; Pim-1/-2/-3 IC50 = 0.4/5.0/1.9 nM with [ATP] = Km; Pim-1/-2/-3 IC50 = 2.6/164/17 nM with [ATP] = 5 mM) wtih high target selectivity (by binding assay with a 442-kinase panel). AZD1208 exhibits selective antiproliferation potency against high Pim-1/-2-expressing AML cultures (GI50 in 72 h = 20 nM/MOLM-16, 60 nM/EOL-1, 300 nM/Kasumi-3, 600 nM/KG-1a, 900 nM/MV4-11) and displays in vivo efficacy against MOLM-16 xenograft tumor growth in mice (by 89% or 100%, respectively, with 10 or 30 mg/kg/day p.o.)
Orally available, potent, selective, ATP-competitive pan-Pim kinase inhibitor against Pim-dependent cancer proliferation in vitro and in vivo.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mehdi Bouhaddou et al.
Cell, 182(3), 685-712 (2020-07-10)
The causative agent of the coronavirus disease 2019 (COVID-19) pandemic, severe acute respiratory syndrome coronavirus 2 (SARS-CoV-2), has infected millions and killed hundreds of thousands of people worldwide, highlighting an urgent need to develop antiviral therapies. Here we present a

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service