Skip to Content
Merck
All Photos(1)

Key Documents

SML2592

Sigma-Aldrich

Arteether

≥95% (HPLC)

Synonym(s):

β-Arteether, (+)-Arteether, (3R,5aS,6R,8aS,9R,10S,12R,12aR)-10-Ethoxydecahydro-3,6,9-trimethyl-3,12-epoxy-12H-pyrano[4,3-j]-1,2-benzodioxepin, Dihydroartemisinin ethyl ether

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€53.60
25 MG
€217.00

€53.60


Estimated to ship on12 April 2025


Request a Bulk Order

Select a Size

Change View
5 MG
€53.60
25 MG
€217.00

About This Item

Empirical Formula (Hill Notation):
C17H28O5
CAS Number:
Molecular Weight:
312.40
MDL number:
UNSPSC Code:
51111800
NACRES:
NA.77

€53.60


Estimated to ship on12 April 2025


Request a Bulk Order

Assay

≥95% (HPLC)

form

powder

optical activity

[α]/D +148 to +165°, c = 1 in chloroform-d

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

O1OC2(O[C@H]3O[C@@H]([C@@H]([C@H]4[C@]31[C@H]([C@@H](CC4)C)CC2)C)OCC)C

InChI

1S/C17H28O5/c1-5-18-14-11(3)13-7-6-10(2)12-8-9-16(4)20-15(19-14)17(12,13)22-21-16/h10-15H,5-9H2,1-4H3/t10-,11-,12+,13+,14+,15-,16?,17-/m1/s1

InChI key

NLYNIRQVMRLPIQ-BVCZABNKSA-N

Biochem/physiol Actions

Antimalarial artemisinin derivative
Arteether is a semisynthetic artemisinin derivative with antimalarial activity. It has been used for the treatment of chloroquine-resistant Plasmodium falciparum malaria and cerebral malaria, and has also been shown to have anticancer activity.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Repr. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service