Skip to Content
Merck
All Photos(1)

Key Documents

SML2591

Sigma-Aldrich

SCH-39166 hydrobromide

≥98% (HPLC)

Synonym(s):

(-)-trans-6,7,7a,8,9,13b-Hexahydro-3-chloro-2-hydroxy-N-methyl- 5H-benzo[d]naptho-(2,1-b)azepine hydrobromide, (6aS,13bR)-11-Chloro-6,6a,7,8,9,13b-hexahydro-7-methyl-5H-benzo[d]naphth[2,1-b]azepin-12-ol hydrobromide, (6aS-trans)-11-Chloro-6,6a,7,8,9,13b-hexahydro-7-methyl-5H-Benzo[d]naphth[2,1-b]azepin-12-ol hydrobromide, Ecopipam hydrobromide, PSYRX 101 hydrobromide, PSYRX-101 hydrobromide, PSYRX101 hydrobromide, SCH 39166 hydrobromide, SCH39166 hydrobromide

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€112.00
25 MG
€448.00

€112.00


Estimated to ship on25 May 2025


Request a Bulk Order

Select a Size

Change View
5 MG
€112.00
25 MG
€448.00

About This Item

Empirical Formula (Hill Notation):
C19H20ClNO·HBr
CAS Number:
Molecular Weight:
394.73
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

€112.00


Estimated to ship on25 May 2025


Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

Biochem/physiol Actions

High-affinity D1/D5 subtype-selective dopamine receptor antagonist with in vitro and in vivo efficacy.
SCH-39166 (ecopipam) is a high-affinity D1/D5 subtype-selective dopamine receptor antagonist (Ki = 1.2 nM/D1, 2.0 nM/D5, 980 nM/D2, 5.52 μM/D4, 80 nM/5-HT, 731 nM/α2a). SCH-39166 is widely employed both in cultures and in animal studies in vivo.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

STOT SE 3

Target Organs

Central nervous system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yang Yang et al.
Molecular psychiatry (2018-12-12)
Dopamine D1 agonists enhance cognition, but the role of different signaling pathways (e.g., cAMP or β-arrestin) is unclear. The current study compared 2-methyldihydrexidine and CY208,243, drugs with different degrees of both D1 intrinsic activity and functional selectivity. 2-Methyldihydrexidine is a
Brendan D Hare et al.
Nature communications, 10(1), 223-223 (2019-01-16)
Impaired function in the medial prefrontal cortex (mPFC) contributes to depression, and the therapeutic response produced by novel rapid-acting antidepressants such as ketamine are mediated by mPFC activity. The mPFC contains multiple types of pyramidal cells, but it is unclear
Meera E Modi et al.
Frontiers in molecular neuroscience, 11, 107-107 (2018-07-05)
Mutations in the SHANK family of genes have been consistently identified in genetic and genomic screens of autism spectrum disorder (ASD). The functional overlap of SHANK with several other ASD-associated genes suggests synaptic dysfunction as a convergent mechanism of pathophysiology
Yunjin Lee et al.
Experimental neurobiology, 27(6), 539-549 (2019-01-15)
Autism spectrum disorder (ASD) is a heterogeneous group of neurobehavioral disorders characterized by the two core domains of behavioral deficits, including sociability deficits and stereotyped repetitive behaviors. It is not clear whether the core symptoms of ASD are produced by
Takeshi Enomoto et al.
Behavioral neuroscience, 132(6), 526-535 (2018-10-10)
Effort-based decision-making paradigms have recently been used to measure motivation in healthy subjects and patients with neuropsychiatric disorders. In the present study, we developed a novel effort-discounting paradigm using a touch-panel system in common marmosets. Marmosets were trained to choose

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service