Skip to Content
Merck
All Photos(1)

Key Documents

SML2590

Sigma-Aldrich

AZD3463

≥98% (HPLC)

Synonym(s):

AZD 3463, AZD-3463, N-(4-(4-Aminopiperidin-1-yl)-2-methoxyphenyl)-5-chloro-4-(1H-indol-3-yl)pyrimidin-2-amine, N-[4-(4-Amino-1-piperidinyl)-2-methoxyphenyl]-5-chloro-4-(1H-indol-3-yl)-2-pyrimidinamine

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€64.50
25 MG
€336.00

€64.50


Check Cart for Availability


Select a Size

Change View
5 MG
€64.50
25 MG
€336.00

About This Item

Empirical Formula (Hill Notation):
C24H25ClN6O
CAS Number:
Molecular Weight:
448.95
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

€64.50


Check Cart for Availability

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

ClC1=CN=C(N=C1C2=CNC3=C2C=CC=C3)NC4=CC=C(C=C4OC)N5CCC(CC5)N

InChI

1S/C24H25ClN6O/c1-32-22-12-16(31-10-8-15(26)9-11-31)6-7-21(22)29-24-28-14-19(25)23(30-24)18-13-27-20-5-3-2-4-17(18)20/h2-7,12-15,27H,8-11,26H2,1H3,(H,28,29,30)

InChI key

GCYIGMXOIWJGBU-UHFFFAOYSA-N

Biochem/physiol Actions

AZD3463 is an orally available, potent receptor tyrosine kinase (RTK) inhibtor against anaplastic lymphoma kinase ALK (Ki = 0.75 nM; IC50 = 15-21 nM with 5FAM-Srctide as substrate & 30 μM ATP), insulin-like growth factor (IGF)-1 receptor (IGF1R), ROS1 (c-Ros), and Trk-A (NTRK1). AZD3463 effectively inhibits multiple crizotinib-resistant cancer growth and displays in vivo efficacy against FLT3-ITD-dependent MOLM-13 acute myeloid leukemia (AML) as well as NGP (wt ALK) & SH-SY5Y (ALK-F1174L) neuroblastoma in mice (15 mg/kg ip.).
Orally available, potent RTK inhibtor against anaplastic lymphoma kinase (ALK), IGF1R, ROS1 and Trk-A (NTRK1) with in vitro and in vivo anticancer efficacy.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service