Skip to Content
Merck
All Photos(1)

Key Documents

SML2517

Sigma-Aldrich

Tanshinone II-A sodium sulfonate

≥95%

Synonym(s):

6,7,8,9,10,11-hexahydro-1,6,6-Trimethyl-10,11-dioxo-phenanthro[1,2-b]furan-2-sulfonic acid sodium salt, Sodium tanshinone IIA sulfonate, Sulfotanshinone sodium II-A, TIIAS

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
€54.50

€54.50


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
€54.50

About This Item

Empirical Formula (Hill Notation):
C19H17NaO6S
CAS Number:
Molecular Weight:
396.39
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

€54.50


Check Cart for Availability

Request a Bulk Order

Assay

≥95%

form

powder

color

white to beige

solubility

H2O: 2 mg/mL, clear (warmed)

storage temp.

2-8°C

SMILES string

CC1(C)CCCC2=C3C(C(OC(S(=O)(O[Na])=O)=C4C)=C4C(C3=O)=O)=CC=C21

InChI

1S/C19H18O6S.Na/c1-9-13-15(20)16(21)14-10-5-4-8-19(2,3)12(10)7-6-11(14)17(13)25-18(9)26(22,23)24;/h6-7H,4-5,8H2,1-3H3,(H,22,23,24);/q;+1/p-1

InChI key

AZEZEAABTDXEHR-UHFFFAOYSA-M

Biochem/physiol Actions

Tanshinone II A sulfonate is a water-soluble derivative of tanshinone II A that exhibits anti-inflammatory properties in rodent models of hepatitis and sepsis. Tanshinone II A sulfonate targets Ca2+-activated potassium channels and carboxylesterase 2. Recent studies indicate that tanshinone II A sulfonate but not tanshinone II A acts as potent negative allosteric modulator of the human purinergic receptor P2X7.
anti-inflammatory, potent negative allosteric modulator of the human purinergic receptor P2X7

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Kaiser et al.
The Journal of pharmacology and experimental therapeutics, 350(3), 531-542 (2014-06-28)
Tanshinone II A sulfonate (TIIAS) was identified as a potent, selective blocker of purinergic receptor P2X7 in a compound library screen. In this study, a detailed characterization of the pharmacologic effects of TIIAS on P2X7 is provided. Because TIIAS is
Jian Wang et al.
American journal of respiratory cell and molecular biology, 48(1), 125-134 (2012-10-16)
Danshen, the dried root of Salvia miltiorrhiza, is widely used in clinics in China for treating various diseases, including cardiovascular diseases. Sodium tanshinone IIA sulfonate (STS), a water-soluble derivative of tanshinone IIA isolated as the major active component from Danshen
Ruijuan Guan et al.
Frontiers in pharmacology, 9, 263-263 (2018-05-17)
Aberrant activation of hypoxia-inducible factor (HIF)-1α is frequently encountered and promotes oxidative stress and inflammation in chronic obstructive pulmonary disease (COPD). The present study investigated whether sodium tanshinone IIA sulfonate (STS), a water-soluble derivative of tanshinone IIA, can mediate its

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service